Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This gene encodes an isoform of the alpha subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This isoform (alpha 2) is expressed in brain, heart and skeletal muscle, and the other isoform (alpha 1) is expressed in brain, placenta, lung, liver, kidney, and pancreas. This gene may be critical in the development of ovarian cancer. [provided by RefSeq, Mar 2014]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eEF1A-2; EF-1-alpha-2; elongation factor 1 A-2; Elongation factor 1-alpha 2; Eukaryotic elongation factor 1 A-2; statin S1; Statin-like protein; Statin-S1
Gene Aliases: DEE33; Eef1a; EEF1A2; EEF1AL; EF-1-alpha-2; EF1A; EIEE33; HS1; MRD38; Ps10; RATPS10; S1; STN; STNL; wasted; wst
UniProt ID: (Human) Q05639, (Mouse) P62631, (Rat) P62632
Entrez Gene ID: (Human) 1917, (Mouse) 13628, (Rat) 24799
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support