What is the strain of E.coli used in One Shot PIR1 Chemically Competent E. coli (Cat. No. C101010)?
The strain of E. coli used is K12.
Find additional tips, troubleshooting help, and resources within our Cloning Support Center.
Find additional tips, troubleshooting help, and resources within our Cloning Support Center.