What is the expected peptide yield for samples prepared with the Click-iT Protein Enrichment Kit, for click chemistry capture of azide-modified proteins (Cat. No. C10416)?
The yield of purified peptides from samples prepared with the Click-iT Protein Enrichment Kit, for click chemistry capture of azide-modified proteins (Cat. No. C10416), strongly depends on the sample type, the incorporation efficiency of the azide analog used, and the amount of lysate added to the resin. In the examples presented in the posters linked below, we were able to recover approximately 25-50 µg of purified peptides from about 0.5 to 8 mg of cell lysate, using approximately 100 µL of resin.
Click chemistry enrichment of cell surface glycoproteins
A universal click chemistry-based enrichment column for the identification of multiple subclasses of PTM-modified proteins and newly synthesized proteins
Click chemistry enrichment of cell surface glycoproteins
A universal click chemistry-based enrichment column for the identification of multiple subclasses of PTM-modified proteins and newly synthesized proteins