Which sample types can I use with the CellEvent Senescence Green Detection Kit?
The CellEvent Senescence Green Detection Kit can be used with various sample types, including fixed cells, adherent cells, and tissue sections.