CellTrace™ BODIPY™ TR Methyl Ester (Lipophilic Counterstain For GFP)
Invitrogen™
CellTrace™ BODIPY™ TR Methyl Ester (Lipophilic Counterstain For GFP)
The red-fluorescent CellTrace™ BODIPY® TR methyl ester is an excellent counterstain for cells and tissues that are expressing GFP. CellTrace™Read more
Have Questions?
Catalog NumberQuantity
C345561 mL
Catalog number C34556
Price (USD)
410.00
Special offer
Online exclusive
Ends: 31-Dec-2026
500.00
Save 90.00
Each
Quantity:
1 mL
Price (USD)
410.00
Special offer
Online exclusive
Ends: 31-Dec-2026
500.00
Save 90.00
Each
The red-fluorescent CellTrace™ BODIPY® TR methyl ester is an excellent counterstain for cells and tissues that are expressing GFP. CellTrace™ BODIPY® TR methyl ester readily permeates cell membranes and selectively stains mitochondria and endomembranous organelles such as endoplasmic reticulum and the Golgi apparatus, but does not appear to localize in the plasma membrane. CellTrace™ BODIPY® TR methyl ester staining is retained after paraformaldehyde fixation.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dye TypeBODIPY Dyes
Product LineBODIPY, CellTrace
Quantity1 mL
Reagent TypeCell Tracker Compounds, Cell Labeling Reagents
Shipping ConditionRoom Temperature
Label TypeOther Labels or Dyes
Product TypeLipophilic Tracer
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.
Frequently asked questions (FAQs)
I am storing the DMSO stock solutions of the CellTrace cell proliferation reagents and am not obtaining a good signal with my cells compared to freshly dissolved dye stock. Why does the product not work after storage?
We provide the CellTrace reagents in small aliquots and strongly recommend discarding any unused DMSO/dye stocks. The CellTrace reagents have acetyl groups to cap the charges on the dyes to make them cell permeant, and succinimidyl ester amine-reactive moiety to allow for covalent attachment to cellular components for long-term retention. Both acetyl groups and succinimidyl esters will readily hydrolyze if any water is present during storage. DMSO is hygroscopic and thus readily absorbs water from the atmosphere. If you must store your dye stocks, you will need to use a good quality, anhydrous DMSO stock that has not been opened often and store the vial within an air-tight container containing some desiccant to keep the DMSO/dye stock solution anhydrous during storage.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
I am using the CellTrace Cell Proliferation reagents and am not obtaining good separation of my cell generation peaks. How can I improve the peak separation?
Here are some tips to obtain uniform staining and a bright, unstimulated parent generation peak:
Dissolve the CellTrace dye stock immediately before use in the DMSO provided in the kit or in good quality, anhydrous DMSO to obtain the best reactivity and cell permeability.
Stain in PBS or other amine-free, protein-free physiological buffer. Do not stain in medium.
Start with a single-cell suspension and gently agitate the cells during staining.
Quickly remove the unbound dye by incubating the cells in ice-cold media for 5 minutes and then wash twice more with pre-warmed media.
Include a dead-cell stain in the assay and gate only on live cells.
Analyze as many cells as possible from each sample.
Use a low flow rate for analysis on hydrodynamic focusing cytometers.
A good staining concentration for the CellTrace dyes is generally within 1-10 µM, but the optimal concentration for a particular cell type will vary. Observe your cells in a stain dilution series to determine the optimal concentration for your cells.
Some cell types may take up dye with a broad staining intensity distribution. If this is the case for your cells, then you will need to do an initial sort of the stained, unstimulated parent cells to select for a narrow peak distribution.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Can I store the stock solutions of the CellTrace reagents and how do you recommend storing them?
We provide the CellTrace reagents in small aliquots and strongly recommend discarding any unused DMSO dye stocks. Some of the CellTrace reagents have diacetate groups to cap the charges on the dyes to make them cell permeant, and some have succinimidyl ester amine-reactive groups for long-term cellular retention. Both diacetates and succinimidyl esters will readily hydrolyze if any water is present during storage. DMSO is very hygroscopic and thus readily absorbs water from the atmosphere. We do not recommend storing stock solutions of CellTrace reagents because storage of the product in solution will inevitably lead to partial or complete loss of reactivity.
How do the CellTrace Cell Proliferation reagents work?
CellTrace Cell Proliferation reagents are all cell-permeant dyes that are cleaved by intracellular esterases to yield highly fluorescent compounds that also covalently bind to cellular amines, attaching the dye to various cellular components and providing a very stable signal. These reagents show little cytotoxicity with minimal observed effects on the proliferative ability of many cells.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
I am trying to assay cell proliferation with a CellTrace stain, and I am not seeing separate peaks for each cell division. How can I optimize this assay?
The key to good generational profiles with CellTrace reagents is starting with cells that are evenly labeled so that they have a tight coefficient of variance (CV) when run at time zero after labeling. If the peak is too broad, the generations will overlap each other and the series of peaks will become a hump. Even labeling can be achieved by starting with a uniform cell population (not a mixture of lymphocytes and granulocytes for example) as staining will be proportional to cell size. Cells are labeled rapidly, so you want to pre-dilute the dye and mix it into your cells rapidly. Be sure that the cells are not sitting in a clump in the bottom of your tube. The easiest way to do this is to make a 2x dye solution (1x = 1-10 µM) and resuspend your cells in a half volume of medium (no serum or BSA). Add the dye to the cells and invert a few times to mix. Gently agitate the cells during staining. Once the dye incubation is over (20 min, 37 degrees C), add serum or BSA (at least 1%) to scavenge any remaining unreacted dye. Spin down cells, wash 1x, and resuspend in complete medium. After a 10-20 min incubation to undergo de-esterification, cells are ready to be set up for whatever treatment you are planning. Be sure to keep a time zero control as you need to know where the first generation ran.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Citations & References (18)
Citations & References
Abstract
Live-cell imaging of rhabdovirus-induced morphological changes in plant nuclear membranes.
Authors:Goodin M, Yelton S, Ghosh D, Mathews S, Lesnaw J
Journal:Mol Plant Microbe Interact
PubMed ID:16042016
'Potato yellow dwarf virus (PYDV) and Sonchus yellow net virus (SYNV) belong to the genus Nucleorhabdovirus. These viruses replicate in nuclei of infected cells and mature virions accumulate in the perinuclear space after budding through the inner nuclear membrane. Infection of transgenic Nicotiana benthamiana 16c plants (which constitutively express green ... More
Fragile X mental retardation protein FMRP binds mRNAs in the nucleus.
Authors:Kim M, Bellini M, Ceman S,
Journal:Mol Cell Biol
PubMed ID:18936162
The fragile X mental retardation protein FMRP is an RNA binding protein that associates with a large collection of mRNAs. Since FMRP was previously shown to be a nucleocytoplasmic shuttling protein, we examined the hypothesis that FMRP binds its cargo mRNAs in the nucleus. The enhanced green fluorescent protein-tagged FMRP ... More
Transcriptional repression mediated by repositioning of genes to the nuclear lamina.
Authors:Reddy KL, Zullo JM, Bertolino E, Singh H,
Journal:Nature
PubMed ID:18272965
Nuclear compartmentalization seems to have an important role in regulating metazoan genes. Although studies on immunoglobulin and other loci have shown a correlation between positioning at the nuclear lamina and gene repression, the functional consequences of this compartmentalization remain untested. We devised an approach for inducible tethering of genes to ... More
Conditional lethality, division defects, membrane involution, and endocytosis in mre and mrd shape mutants of Escherichia coli.
Authors:Bendezú FO, de Boer PA,
Journal:J Bacteriol
PubMed ID:17993535
Maintenance of rod shape in Escherichia coli requires the shape proteins MreB, MreC, MreD, MrdA (PBP2), and MrdB (RodA). How loss of the Mre proteins affects E. coli viability has been unclear. We generated Mre and Mrd depletion strains under conditions that minimize selective pressure for undefined suppressors and found ... More
POLKADOTS are foci of functional interactions in T-Cell receptor-mediated signaling to NF-kappaB.
Authors:Rossman JS, Stoicheva NG, Langel FD, Patterson GH, Lippincott-Schwartz J, Schaefer BC
Journal:Mol Biol Cell
PubMed ID:16495340
Stimulation of the T-cell receptor (TCR) results in the activation of several transcription factors, including NF-kappaB, that are crucial for T-cell proliferation and gain of effector functions. On TCR engagement, several proteins within the TCR-directed NF-kappaB signaling pathway undergo dynamic spatial redistribution, but the significance of these redistribution events is ... More