CyQUANT™ Direct Cell Proliferation Assay, 10 microplates kit - FAQs
CyQUANT™ Direct Cell Proliferation Assay, 10 microplates kit - FAQs
View additional product information for CyQUANT™ Cell Proliferation Assays - FAQs (C35012, C35013, C35011, C7027, C7026, C35006, C35007)
7 product FAQs found
MTT cell proliferation plate assays require cellular metabolism to modify the reagent and thus, only live cells will be counted. Is this true for CyQUANT Direct Cell Proliferation Assay, too?
This is true for MTT (as well as XTT, AlamarBlue Cell Viability Reagent, and PrestoBlue Cell Viability Reagent). CyQUANT Direct will also only count live cells, but for a different reason. The dye in it is a green-fluorescent nucleic acid stain, which will bind to DNA in all cells, live or dead, without the need for cellular metabolism. However, there is a non-cell-permeable quenching reagent in the kit which will both quench extracellular fluorescence (and thus this is a no-wash assay) AND will quench the fluorescence in dead cells (but not live cells).
MTT cell proliferation assays require cellular metabolism to turn over the reagent, and thus only live cells will be counted. Is this true for CyQUANT Direct as well?
This is true for MTT (as well as XTT, AlamarBlue Cell Viability Reagent, and PrestoBlue Cell Viability Reagent). CyQUANT Direct will also only count live cells, but for a different reason. The dye is a green-fluorescent nucleic acid stain, which will bind to DNA in all cells, live or dead, without the need for cellular metabolism. However, there is a cell impermeant quenching reagent in the kit which will quench both extracellular fluorescence (and thus this is a no-wash assay) and the fluorescence in dead cells (but not live cells).
What volumes should I use to make a 10X stock solution for the CyQUANT Direct Cell Proliferation Assay (Cat. No. C35011, C35012)?
To make a 10X stock solution for the CyQUANT Direct Cell Proliferation Assay (Cat. No. C35011, C35012), we suggest using the following volumes:
1.1 mL         PBS
25 µL         CyQUANT Direct nucleic acid stain (Component A)
125 µL         CyQUANT Direct background suppressor I (Component B)
Are the components of the CyQUANT Direct Red Cell Proliferation Assay and CyQuant Direct Cell Proliferation Assay kits interchangeable?
No. The components of the CyQUANT Direct Red Cell Proliferation Assay and CyQuant Direct Cell Proliferation Assay kits are not interchangeable.
What is the difference between the CyQUANT Direct Red Cell Proliferation Assay (Cat. Nos. C35013, C35014) and the green fluorescent CyQuant Direct Cell Proliferation Assay (Cat Nos. C35011, C35012)?
The two kits utilize different dyes and background suppressors, but they function in the same way and provide similar results as can be seen from Figures 1 and 2 in the CyQUANT Direct Red Cell Proliferation Assay product sheet (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018644_CyQUANT_direct_red_assay_PI.pdf). Dyes from both kits are nucleic acid stains and the background suppressors are cell impermeant reagents that quench extracellular fluorescence. They are usable within the same range (100 to 20,000 cells/well in a 96-well format) and in the same workflow.
Do you offer any products to measure neuronal cell health?
PrestoBlue Cell Viability Stain and CyQUANT Cell Proliferation Assay Kit can be used. We also offer a Neurite Outgrowth Staining Kit (Cat. No. A15001). More information about our different assays for neuronal cell health can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/neuroscience/neuronal-cell-health-assays.html).
Can other cell viability reagents be used with AlgiMatrix Matrix?
Other than the Invitrogen alamarBlue Cell Viability Reagent, AlgiMatrix Matrix is also compatible with Invitrogen PrestoBlue Cell Viability Reagent and Invitrogen CyQuant Direct Cell Proliferation Assay. The protocol is almost the same as the 2D cell culture, but it needs the blank AlgiMatrix Matrix as a blank control.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.