Cascade Blue™ hydrazide, Trisodium Salt - FAQs
Cascade Blue™ hydrazide, Trisodium Salt - FAQs
View additional product information for Cascade Blue™ hydrazide, Trisodium Salt - FAQs (C687)
1 product FAQs found
What is the recommended concentration for preparation of Cascade Blue hydrazide, Trisodium Salt (Cat. No. C687) stock solution?
We recommend preparing a 1% w/v stock solution in sterile water. Please note that the dye can aggregate in solution over time.