Can I verify that my NanoDrop One/Onec Spectrophotometer is calibrated and running accurately?
Yes. While most users will never need to recalibrate their NanoDrop One/Onec Spectrophotometer, we recommend verifying instrument performance using the NanoDrop PV-1 Performance Verification Solution every 6 months. PV-1 is available from the Thermo Fisher website.