NanoDrop™ Reagents
Thermo Scientific™

NanoDrop™ Reagents

Certified standards for verifying instrument performance to factory specifications are available separately.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
CHEM-PV-1PV-1 Performance Verification
CHEM-CF-1CF-1 Calibration Check Fluid
CHEM-CF-8CF-8 Calibration Check Kit
CHEM-PR1-KITND PR-1 Reconditioning Kit
CHEM-PV-8PV-8 Performance Verification
Catalog number CHEM-PV-1
Price (USD)
149.00
Each
Type:
PV-1 Performance Verification
Price (USD)
149.00
Each
Certified standards for verifying instrument performance to factory specifications are available separately.

CHEM-PV-1 - PV-1 Performance Verification Solution

For use with NanoDrop Ultra Spectrophotometers and Fluorometers, NanoDrop One/OneC and NanoDrop Lite Plus
Solution with certified absorbance values.
Use to create a record of correct photometric performance.
Intended for single use only.
 

CHEM-PV-8 - PV-8 Performance Verification Kit

For use with NanoDrop Eight
Solution with certified absorbance values.
Use to create a record of correct photometric performance.
Intended for single use only.
 

CHEM-CF-1 - CF-1 Calibration Fluid

For use with NanoDrop 1000, NanoDrop 2000/2000c, NanoDrop Lite
Solution with certified absorbance values.
Use to create a record of correct photometric performance.
Intended for single use only.
 

CHEM-CF-8 - CF-8 Calibration Check Kit

For use with NanoDrop 8000
Solution with certified absorbance values.
Use to create a record of correct photometric performance.
Intended for single use only.
 

CHEM-PR1-KIT - PR-1 Reconditioning Kit

For use with all NanoDrop models
This kit is useful for reconditioning the sample measurement pedestals to a hydrophobic state if they become “unconditioned” (see user manual for more information). The PR-1 kit consists of a small screw cap vial of specially formulated compound and a supply of 25 convenient applicators.

Specifications
For Use With (Equipment)NanoDrop Ultra, NanoDrop One and NanoDrop OneC spectrophotometers
Storage RequirementsStore at room temperature
TypePV-1 Performance Verification
Unit SizeEach

Frequently asked questions (FAQs)

Can I verify that my NanoDrop One/Onec Spectrophotometer is calibrated and running accurately?

Yes. While most users will never need to recalibrate their NanoDrop One/Onec Spectrophotometer, we recommend verifying instrument performance using the NanoDrop PV-1 Performance Verification Solution every 6 months. PV-1 is available from the Thermo Fisher website.