Accessories for Cimarec+™ Stirrer Series
Accessories for Cimarec+™ Stirrer Series
Get precise stirring control for your routine protocols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
CIC0001446Transparent shield for 10x10
CIC0001448PT1000 External probe 0-400C
CIC0001444Transparent shield for 4x4
CIC0001445Transparent shield for 7x7
CIC0001830Supporting Rod
CIC0001831Clamp and holder
Catalog number CIC0001446
Price (MXN)
-
Type:
Transparent shield for 10x10
These accessories are for use with the Cimarec+™ series digital stirrers. Our new Thermo Scientific™ Cimarec+™ series digital stirrers offer precise stirring control for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operation.
Advanced technology
  • Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strong magnetic coupling
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for stirring
  • Raised display design protects electronics from spills
  • Optional splash guard protection shield
  • Variable sizes for different sample volumes
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
TypeTransparent shield for 10x10
DisplayLED
Unit SizeEach