CellInsight™ CX5 High Content Screening (HCS) Platform - FAQs
CellInsight™ CX5 High Content Screening (HCS) Platform - FAQs
View additional product information for CellInsight™ CX5 High Content Screening (HCS) Platform - FAQs (CX51110)
2 product FAQs found
Who do I contact if I have issues with my CellInsight systems or ArrayScan System?
Contact Instrument Customer Services for any issues regarding the CellInsight systems and ArrayScan System. They can be reached at instrumentservices@thermofisher.com.
For high-content analysis (HCA)/high-content screening (HCS), what are the main differences between the CellInsight CX5, CX7, and ArrayScan systems?
The basic differences are in the number of channels for excitation (5 versus 7), the ability to image in brightfield, widefield, and confocal, and the number of positions available on the objective turret. The CellInsight CX5 System is the most basic HCS system with illumination in 5 channels, imaging in brightfield and widefield, and a 1-position objective turret. The CellInsight CX7 System and ArrayScan System provide illumination in 7 channels, imaging in brightfield, widefield, and confocal, and either a 3-position (CX7) or 4-position objective turret (ArrayScan System). The ArrayScan System has the added option of live-cell imaging. For more information, go here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cellular-imaging/high-content-screening.html).