BODIPY™ 650/665-X NHS Ester (Succinimidyl Ester)
Invitrogen™

BODIPY™ 650/665-X NHS Ester (Succinimidyl Ester)

BODIPY™ 650/665-X dye is bright, far-red fluorescent dye with similar excitation and emission to Cy5 or Alexa Fluor™ 647. ItRead more
Have Questions?
Catalog NumberQuantity
D10001
also known as D-10001
5 mg
Catalog number D10001
also known as D-10001
Price (USD)
430.65
Online exclusive
574.00
Save 143.35
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
430.65
Online exclusive
574.00
Save 143.35
Each
BODIPY™ 650/665-X dye is bright, far-red fluorescent dye with similar excitation and emission to Cy5 or Alexa Fluor™ 647. It has a high extinction coefficient and fluorescence quantum yield and is relatively insensitive to solvent polarity and pH change. In contrast to the highly water soluble fluorophores Alexa Fluor™ 488 dye and fluorescein (FITC), BODIPY™ dyes have unique hydrophobic properties ideal for staining lipids, membranes, and other lipophilic compounds. BODIPY™ 650/665-X dye has a relatively long excited-state lifetime (typically 5 nanoseconds or longer), which is useful for fluorescence polarization-based assays and a large two-photon cross-section for multiphoton excitation. In addition to reactive dye formulations, we offer BODIPY™ 650/665-X dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection.

The NHS ester (or succinimidyl ester) of BODIPY™ 650/665-X is the most popular tool for conjugating the dye to a protein or antibody. NHS esters can be used to label the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting BODIPY™ 650/665-X conjugates exhibit bright fluorescence, narrow emission bandwidths, and relatively long excited-state lifetimes, which can be useful for fluorescence polarization assays and two-photon excitation (TPE) microscopy.

This reactive dye contains a seven-atom aminohexanoyl ('X') spacer between the fluorophore and the NHS ester group. This spacer helps to separate the fluorophore from its point of attachment, potentially reducing the interaction of the fluorophore with the biomolecule to which it is conjugated.

Detailed information about this BODIPY™ 650/665-X NHS ester:

Fluorophore label: BODIPY™ 650/665-X dye
Reactive group: NHS ester (succinimidyl ester)
Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
Ex/Em of the conjugate: 646/660 nm
Extinction coefficient: 102,000 cm-1M-1
Molecular weight: 643.45

Typical Conjugation Reaction
Amine-reactive reagents can be conjugated with virtually any protein or peptide; the provided protocol is optimized for IgG antibodies. The reaction can be scaled for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The BODIPY™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO), and the reaction is carried out in 0.1-0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification
Labeled antibodies are typically separated from free BODIPY™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration medium with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Emission660 nm
Excitation646 nm
Label or DyeBODIPY™ 650⁄665
Product TypeDye
Quantity5 mg
Reactive MoietyActive Ester, Succinimidyl Ester
Shipping ConditionRoom Temperature
Label TypeBODIPY Dyes
Product LineBODIPY
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (6)

Citations & References
Abstract
Synthesis and pharmacological characterization of novel fluorescent histamine H2-receptor ligands derived from aminopotentidine.
Authors:Xie SX, Petrache G, Schneider E, Ye QZ, Bernhardt G, Seifert R, Buschauer A,
Journal:Bioorg Med Chem Lett
PubMed ID:16730977
'In an effort to develop a non-radioactive alternative to the [3H]tiotidine and [125I]iodoaminopotentidine binding assays for the histamine H2-receptor (H2R), primary amines related to aminopotentidine were prepared and coupled with the succinimidyl esters of the bulky fluorescent dyes S0536 and BODIPY 650/665-X. The primary amines exhibited different degrees of antagonistic ... More
Multicolor in vitro translation.
Authors:Traverso G, Diehl F, Hurst R, Shuber A, Whitney D, Johnson C, Levin B, Kinzler KW, Vogelstein B
Journal:Nat Biotechnol
PubMed ID:12910244
'In vitro translation is a widely used tool for both analytical and preparative purposes. For analytical purposes, small amounts of proteins are synthesized and visualized by detection of labeled amino acids incorporated during translation. The original strategy of incorporating radioactively labeled amino acids, such as [35S]methionine or [14C]leucine, has been ... More
Glycolipid trafficking in Drosophila undergoes pathway switching in response to aberrant cholesterol levels.
Authors:Hortsch R, Lee E, Erathodiyil N, Hebbar S, Steinert S, Lee JY, Chua DS, Kraut R,
Journal:Mol Biol Cell
PubMed ID:20053687
In lipid storage diseases, the intracellular trafficking of sphingolipids is altered by conditions of aberrant cholesterol accumulation. Drosophila has been used recently to model lipid storage diseases, but the effects of sterol accumulation on sphingolipid trafficking are not known in the fly, and the trafficking of sphingolipids in general has ... More
Single-molecule detection and identification of multiple species by multiparameter fluorescence detection.
Authors:Widengren J, Kudryavtsev V, Antonik M, Berger S, Gerken M, Seidel CA
Journal:Anal Chem
PubMed ID:16536444
Two general strategies are introduced to identify and quantify single molecules in dilute solutions by employing a spectroscopic method for data registration and specific burst analysis, denoted multiparameter fluorescence detection (MFD). MFD uses pulsed excitation and time-correlated single-photon counting to simultaneously monitor the evolution of the eight-dimensional fluorescence information (fundamental ... More
2,2'-thiodiethanol: a new water soluble mounting medium for high resolution optical microscopy.
Authors:Staudt T, Lang MC, Medda R, Engelhardt J, Hell SW
Journal:Microsc Res Tech
PubMed ID:17131355
The use of high numerical aperture immersion lenses in optical microscopy is compromised by spherical aberrations induced by the refractive index mismatch between the immersion system and the embedding medium of the sample. Especially when imaging >10 micro m deep into the specimen, the refractive index mismatch results in a ... More