EnzChek™ Direct Phospholipase C Assay Kit - FAQs
EnzChek™ Direct Phospholipase C Assay Kit - FAQs
View additional product information for EnzChek™ Direct Phospholipase C Assay Kit - FAQs (E10215)
1 product FAQs found
What is the excitation/emission maxima of the reaction product when using the EnzChek Direct Phospholipase C Assay kit?
The fluorescence excitation/emission maxima is 509/516 nm.