EnzChek™ Direct Phospholipase C Assay Kit
Invitrogen™

EnzChek™ Direct Phospholipase C Assay Kit

Monitors PC-PLC activity

Have Questions?
Catalog NumberQuantity
E102151 Kit
Catalog number E10215
Price (USD)
387.00
Each
Quantity:
1 Kit
Price (USD)
387.00
Each

This kit provides continuous monitoring of phosphotidyl choline specific phospholipase C (PLC) activity in microplate based biochemical assays. The assay uses a PCL selective fluorogenic substrate that emits a bright green emission upon cleavage.

The EnzChek™ Direct Phospholipase C Assay Kit provides a simple and robust method for monitoring PC-PLC activity. Each kit provides enough reagents for 2 microplates, using 200 μl volumes in 96 well format. PC-PLC plays a crucial role in many cell signaling pathways involved in apoptosis and cell survival, as well as diseases ranging from cancer to HIV1-7. The assay uses a glycerophospho-ethanolamine with a dye-labeled sn-2 acyl chain as a substrate for PC-PLC. Substrate cleavage by PC-PLC before the phosphate releases the dye-labeled diacylglycerol, which produces a positive fluorescence signal that may be measured continuously. The reaction product has absorption and fluorescence emission maxima of 509 nm and 516 nm, respectively. Using purified enzyme from Bacillus cereus, the assay can detect as little as 10 mU/mL PC-PLC after one hour at room temperature. The kit has been proven useful for characterizing PC-PLC inhibition, and since it offers a direct measurement, the potential for false positives in a compound screen is eliminated.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeBODIPY
Emission Wavelength516 nm
Excitation Wavelength509 nm
Filter TypeFITC (Fluorescein) Filter
Quantity1 Kit
Shipping ConditionRoom Temperature
Substrate PropertiesLipid-Based Substrate, Chemical Substrate
Substrate TypePhospholipase Substrate
Target EnzymePhosphatidylcholine-Specific Phospholipase C
ColorGreen
For Use With (Application)Enzyme and Metabolic Activity
For Use With (Equipment)Microplate Reader
Product LineEnzChek
Product TypeEnzChek Assay Kit
Unit SizeEach
Contents & Storage
I kit Store kit at -<20°, protected from light, desiccated

Frequently asked questions (FAQs)

What is the excitation/emission maxima of the reaction product when using the EnzChek Direct Phospholipase C Assay kit?

The fluorescence excitation/emission maxima is 509/516 nm.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (4)

Citations & References
Abstract
Membrane Chaperone SecDF Plays a Role in the Secretion of Listeria monocytogenes Major Virulence Factors.
Authors:Burg-Golani T, Pozniak Y, Rabinovich L, Sigal N, Nir Paz R, Herskovits AA,
Journal:
PubMed ID:24056100
'Listeria monocytogenes is a Gram-positive human intracellular pathogen that infects diverse mammalian cells. Upon invasion, L. monocytogenes secretes multiple virulence factors that target host cellular processes and promote infection. It has been presumed, but was not empirically established, that the Sec translocation system is the primary mediator of this secretion. ... More
Lipid peroxidation is essential for phospholipase C activity and the inositol-trisphosphate-related Ca2+ signal.
Authors:Domijan AM, Kovac S, Abramov AY,
Journal:
PubMed ID:24198393
'Reactive oxygen species (ROS) are produced in enzymatic and non-enzymatic reactions and have important roles in cell signalling but also detrimental effects. ROS-induced damage has been implicated in a number of neurological diseases; however, antioxidant therapies targeting brain diseases have been unsuccessful. Such failure might be related to inhibition of ... More
Short- and Long-Term Regulation of Intestinal Na+/H+ Exchange Activity Associated with TLR2 Receptor Activation Is Independent of Nuclear Factor-?B Signaling.
Authors:Cabral JM, Soares-da-Silva P, Magro F,
Journal:
PubMed ID:23845891
'Type 2 Toll-like receptors (TLR2s) are expressed in cell membranes and recognize a wide range of pathogen-associated molecular patterns derived from bacteria, such as lipoteichoic acid (LTA). The aim of this study was to evaluate the effect of TLR2 activation by LTA on the activity of type 1 Na(+)/H(+) exchanger ... More
Lipid-modifying enzymes in human tear fluid and corneal epithelial stress response.
Authors:Robciuc A, Rantamäki AH, Jauhiainen M, Holopainen JM,
Journal:
PubMed ID:24302584
Since homeostasis at the ocular surface requires a delicate balance between numerous factors, and the external environment contributes as an unpredictable component, we aimed to understand the role that various lipids and their regulators have in the complex process that maintains a healthy corneal surface. Through basic proteomics, we tested ... More