EnzChek™ Phospholipase A1 Assay Kit - FAQs
EnzChek™ Phospholipase A1 Assay Kit - FAQs
View additional product information for EnzChek™ Phospholipase A1 Assay Kit - FAQs (E10219)
1 product FAQs found
What is the LoD for the EnzChek Phospholipase A1 Assay Kit (Cat. No. E10219)?
The EnzChek Phospholipase A1 Assay Kit (Cat. No. E10219) can detect PLA1 at 0.04 U/mL or lower.