EnzChek™ Phosphatase Assay Kit - FAQs
EnzChek™ Phosphatase Assay Kit - FAQs
View additional product information for EnzChek™ Phosphatase Assay Kit - FAQs (E12020)
1 product FAQs found
What is the reaction product excitation/emission maxima when using the EnzChek Phosphatase Assay kit?
The reaction product has excitation/emission maxima of approximately 358/455 nm. Fluorescence can be measured in a fluorescence microplate reader or a standard fluorometer.