Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor™ 488 Streptavidin (Alexa Fluor™ 488 EGF complex) - FAQs
Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor™ 488 Streptavidin (Alexa Fluor™ 488 EGF complex) - FAQs
View additional product information for Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor™ 488 Streptavidin (Alexa Fluor™ 488 EGF complex) - FAQs (E13345)
1 product FAQs found
What is the molecular weight of Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor 488 Streptavidin (Alexa Fluor 488 EGF complex) (Cat. No. E13345)?
The Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor 488 Streptavidin (Alexa Fluor 488 EGF complex) (Cat. No. E13345) has an approximate MW of ~60 kDa.
Find additional tips, troubleshooting help, and resources within our Labeling Chemistry Support Center.