EnzChek™ Ultra Amylase Assay Kit - FAQs
EnzChek™ Ultra Amylase Assay Kit - FAQs
View additional product information for EnzChek™ Ultra Amylase Assay Kit - FAQs (E33651)
1 product FAQs found
What is the excitation/emission maxima of the digestion product from EnzChek Ultra Amylase Assay Kit?
The excitation/emission is 502/512nm.