ELF™ 97 Phosphatase Substrate (ELF™ 97 Phosphate), 0.2 μm Filtered - FAQs
ELF™ 97 Phosphatase Substrate (ELF™ 97 Phosphate), 0.2 μm Filtered - FAQs
View additional product information for ELF™ 97 Phosphatase Substrate (ELF™ 97 Phosphate), 0.2 μm Filtered - FAQs (E6588)
2 product FAQs found
Can the ELF 97 reagent be applied to live cells?
ELF 97 substrate is cell impermeant. It may be used on live cells that exhibit phosphatase activity on the surface of the cells, but not intracellular phosphatase activity in live cells. The alternative product for live-cell detection of phosphatase activity is the cell-permeable Alkaline Phosphatase Live Stain (Cat. No. A14353).
Can ELF 97 stained cell/tissue samples be multiplexed with antibody labeling?
No. The ELF 97 reagent does not covalently attach to any cellular components and may be washed away with any subsequent antibody labeling.