EnzChek™ Pyrophosphate Assay Kit - FAQs
EnzChek™ Pyrophosphate Assay Kit - FAQs
View additional product information for EnzChek™ Pyrophosphate Assay Kit - FAQs (E6645)
1 product FAQs found
What is the sensitivity limit of the EnzCheck Pyrophosphate Assay kit?
The sensitivity limit is 1 mM pyrophosphate (approximately 0.2 µg/mL).