FastAP Thermosensitive Alkaline Phosphatase (1 U/μL), 300 units - FAQs
FastAP Thermosensitive Alkaline Phosphatase (1 U/μL), 300 units - FAQs
View additional product information for FastAP Thermosensitive Alkaline Phosphatase (1 U/μL) - FAQs (EF0654, EF0652, EF0651)
2 product FAQs found
Do I need to remove the FastAP Thermosensitive Alkaline Phosphatase prior to ligation?
No, removal is not necessary. The enzyme can be inactivated in 5 mins at 75 degrees C.
How long does the dephosphorylation take when using your FastAP Thermosensitive Alkaline Phosphatase?
Dephosphorylation takes place in approximately 10 mins at 37 degrees C.