T4 Polynucleotide Kinase (10 U/μL), 500 units - FAQs
View additional product information for T4 Polynucleotide Kinase (10 U/μL) - FAQs (EK0032, EK0031)
1 product FAQs found
Can I use Thermo Scientific T4 Polynucleotide Kinase to directly label 5′-termini of nucleic acids with phosphate?
T4 PNK can label nucleic acids with 5′-phosphate in the exchange reaction using reaction buffer B. However, for efficient labeling we recommend to remove the 5′-phosphate groups of DNA/RNA (e.g. by using Thermo Scientific FastAP, Cat. Nos. EF0651/2/4) and then phosphorylate in the forward reaction using T4 PNK with the buffer A.