Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The Mouse Lymphotactin/XCL1 ELISA quantitates Ms Lymphotactin in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms Lymphotactin.
Principle of the method
The Mouse Lymphotactin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Lymphotactin, otherwise known as XCL1, is a member of the C-chemokine family that uniquely acts as a chemoattractant for lymphocytes, but not monocytes or neutrophils, suggesting an important regulatory role in lymphocyte trafficking and inflammation. Lymphotactin, shown to signal through chemokine XC receptor 1 (XCR1), is detected at the highest level in the spleen and at low levels in the lungs, small intestine and colon, and is abundantly and selectively expressed by activated T cells, in particular CD8+ T cells, and is also produced by NK cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : ATAC, Lptn, LTN, SCM-1, SCM-1a, Scyc1
Gene ID : (Mouse) 16963
Gene symbol : Xcl1
Protein Aliases : c motif chemokine 1, C motif chemokine 1, Cytokine SCM-1, Lymphotaxin, Small-inducible cytokine C1, SCM1alpha, SCM-1alpha, SCM1beta, SCM-1beta, SCM-1-beta, chemokine XCL1, cytokine SCM-1, lymphotaxin, small inducible cytokine subfamily C, member 1 (lymphotactin), small-inducible cytokine C1, unnamed protein product
UniProt ID (Mouse) P47993
If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support