T7 RNA Polymerase (20 U/μL), 5,000 units - FAQs
View additional product information for T7 RNA Polymerase (20 U/μL) - FAQs (EP0111, EP0112)
2 product FAQs found
What is the advantage of using TranscriptAid T7 High Yield Transcription Kit over Thermo Scientific T7 RNA Polymerase with a standard transcription buffer?
TranscriptAid T7 High Yield Transcription Kit is designed to produce both long and short transcripts for applications that require large yields of RNA (milligram amounts). The kit can achieve 10 times greater amount than from conventional in vitro transcription reactions with T7 RNA polymerase due to its unique proprietary enzyme and buffer formulations. Please note that TranscriptAid T7 High Yield Transcription Kit is not recommended for generation of radioactively labelled RNA. Due to large quantities of RNA synthesized with the kit, generation of high specific activity radiolabeled probes would require prohibitively large amounts of radiolabeled nucleotide.
Does the T7 RNA Polymerase (20 U/μl) (Cat. No. EP0111) contain a His-Tag which could be used for purification or conjugation?
The T7 RNA Polymerase (20 U/μl) (Cat. No. EP0111) does not contain a His-Tag, and currently none of our T7 RNA polymerases have a tag available for purification/conjugation.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.