Terminal Deoxynucleotidyl Transferase (20 U/μL), 2,500 units - FAQs
Terminal Deoxynucleotidyl Transferase (20 U/μL), 2,500 units - FAQs
View additional product information for Terminal Deoxynucleotidyl Transferase (20 U/μL) - FAQs (EP0162, EP0161)
1 product FAQs found
Is the Terminal Deoxynucleotidyl Transferase (TdT) within the Pierce Biotin 3' End DNA Labeling Kit (Cat. No. 89818) available separately?
Yes, but not the specific one from that same kit. We have the following comparable Terminal Deoxynucleotidyl Transferases (Cat. No. 10533065, 10533073 or EP0161, EP0162).
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Gel Electrophoresis and Blotting Support Center.