CellLight™ FUCCI Cell Cycle Indicator
Invitrogen™

CellLight™ FUCCI Cell Cycle Indicator

CellLight™ FUCCI Cell Cycle Indicator is a fixable live-cell imaging reagent that works as a cell cycle dye alternative, labeling nuclei red in G1, green in S/G2/M, and yellow at G1/S. Single-vector BacMam delivery and mKate support imaging with standard GFP and Texas Red optics.
Have Questions?
Catalog NumberQuantity
F123451 mL
Catalog number F12345
Price (USD)
495.00
Each
Quantity:
1 mL
Price (USD)
495.00
Each

CellLight™ FUCCI Cell Cycle Indicator is a fixable live-cell imaging reagent that works as a cell cycle dye alternative, labeling nuclei red in G1, green in S/G2/M, and yellow at G1/S. The reagent combines GFP-geminin and mKate-Cdt1 in a single BacMam vector, co-tranducing both reporters while supporting imaging with standard GFP and Texas Red filter sets. It is added directly to cells in complete medium, typically read after 16 to 24 hours, and can be carried into fixation and permeabilization workflows for downstream staining or endpoint phenotyping.

A single BacMam particle co-tranduces GFP-geminin and mKate-Cdt1 from a non-replicating baculovirus-based construct supporting equal delivery of both fluorescent proteins with a single transduction.

CellLight™ FUCCI supports imaging with standard GFP and Texas Red filter sets and excels for timelapse imaging for longitudinal or perturbation studies.

As a transduction reagent the BacMam vector has an excellent safety profile and offers a transient approach in mammalian cells because the baculovirus does not replicate in mammalian hosts.

Features

  • Added directly to cells in complete culture medium, with fluorescence typically assessed after 16 to 24 hours.
  • Compatible with standard fixation and permeabilization workflows, supporting endpoint antibody staining or other fluorescent markers after live-cell imaging.
  • Fits microscopy and high-content imaging workflows used to assess proliferation, cell-cycle perturbation, or population shifts after drug, siRNA, or other experimental treatment.

Transient episomal expression supports short-term studies and with optional continuous culture use when sustained labeling is needed. Optimizing from a starting range of 10 to 100 particles per cell for standard mammalian cultured cells with noted lower performance in hematopoietic lineages.

The CellLight™ FUCCI workflow is compatible with routine cell culture with samples moving into downstream staining with antibodies, landmark dyes, or other endpoint markers after live-cell imaging.

For Research Use Only.
Specifications
Concentration1 x 108 Particles/mL
Detection MethodFluorescence Microscopy
EmissionGFP: 520 nm, mKate: 625 nm
ExcitationGFP: 485 nm, mKate: 588 nm
For Use With (Application)Cell Cycle Analysis, Fluorescence Microscopy, Live Cell Microscopy, High Content Imaging
For Use With (Equipment)Flow Cytometer, Countess Cell Counter
FormatVial
FormulationAqueous Concentrate
Product TypeCell Cycle Indicator
Quantity1 mL
Reagent TypeBacMam
SynonymFUCCI
TargetCell Cycle Phases
ColorGreen, Red
Label TypeFluorescent Protein
Product LineCellLight
Unit SizeEach
Contents & Storage
•1 Vial
•Store at 2°C to 8°C
•Protect from light
•Do not freeze

Frequently asked questions (FAQs)

Can BacMam reagents be handled using standard BSL-1 laboratory practices?

Yes. Baculoviruses are insect viruses that do not replicate in mammalian cells and are commonly handled using standard BSL-1 laboratory practices and personal protective equipment appropriate for routine mammalian cell culture work.

Can cells be transduced multiple times with BacMam reagents?

Yes. Because BacMam transduction is generally well tolerated, additional BacMam reagent can be added after several days if higher expression levels are needed or if a different BacMam-based assay will be performed. BacMam reagents may also remain continuously present in the culture medium to help maintain expression.

How can I optimize BacMam transduction in difficult-to-transduce cells?

BacMam transduction can often be improved by optimizing the particles per cell (PPC) ratio, incubation volume, incubation time, temperature, and cell density. For adherent cells, we recommend a confluence of 70% or lower at the time of transduction. After PPC optimization, adjusting transduction volume is often the next most effective parameter for improving expression levels. The BacMam Enhancer Kit (Cat. No. B10107) may also help increase expression in some cell types.

Can transduced cells be cryopreserved without reducing BacMam expression levels?

Our data show that BacMam-transduced cells can be stored in liquid nitrogen for several months without substantially reducing transgene expression levels after recovery.

Does BacMam transduction affect cell viability or cell health?

BacMam transduction is generally well tolerated, even at high viral particle-to-cell ratios greater than 1,000. However, some BacMam reagents may produce cytotoxic effects at high expression levels depending on the transgene being delivered. High expression of organelle-targeted transgenes may also affect subcellular localization. We recommend optimizing particles per cell (PPC) for each BacMam reagent and cell line combination to balance expression levels and cell health.