Fluo-4, Pentapotassium Salt, cell impermeant - FAQs
Fluo-4, Pentapotassium Salt, cell impermeant - FAQs
View additional product information for Fluo-4, Pentapotassium Salt, cell impermeant - FAQs (F14200)
1 product FAQs found
Can I use Fluo-4, Pentapotassium Salt, cell impermeant (Cat. No. F14200) in low pH conditions (pH 4-5)?
No. pH affects the ability of Fluo-4, Pentapotassium Salt, cell impermeant (Cat. No. F14200) to bind calcium and the fluorescence intensity. The Fluo-4 dye drops in fluorescence below pH 5.