Fluo-5F, AM, cell permeant - Special Packaging
Invitrogen™

Fluo-5F, AM, cell permeant - Special Packaging

Analog of fluo-4 with lower Ca2+-binding affinity, making it suitable for detecting intracellular calcium levels in 1μM to 1 mM range that would saturate response of fluo-3 and fluo-4

Have Questions?
Catalog NumberQuantity
F1422210 x 50 μg
Catalog number F14222
Price (USD)
524.00
Each
Quantity:
10 x 50 μg
Request bulk or custom format
Price (USD)
524.00
Each

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. Fluo-5F, fluo-5N, and fluo-4ff are analogs of fluo-4 with lower Ca2+-binding affinity, making them suitable for detecting intracellular calcium levels in the 1 μM to 1 mM range that would saturate the response of fluo-3 and fluo-4. Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells. These indicators are compatible with excitation at 488 nm by argon-ion laser sources, making them useful for confocal microscopy, flow cytometry, and microplate screening applications.

Calcium Indicator (AM Ester) Specifications:

  • Label (Ex/Em of Ca2+–bound form): Fluo-5F (494/516 nm)
  • Fluorescence intensity increase upon binding Ca2+: >100 fold
  • Kd for Ca2+ in buffer: ∼2.3 μM
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength

Using TPEN to Control Heavy Metal Cations

In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+. Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator (Cat. No. TPEN).

More Choices for Fluorescent Calcium Indicators

We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.

For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeFluo-5F
Emission Wavelength516 nm
Excitation Wavelength494 nm
Quantity10 x 50 μg
Shipping ConditionRoom Temperature
Emission Wavelength Range510 to 525 nm
Excitation Wavelength Range480 to 495 nm
For Use With (Application)Calcium Indicator
For Use With (Equipment)Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager
Product TypeCalcium Indicator
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

I am doing calcium flux imaging with your Fura-2 calibration kit, but am seeing a large variability in ratio in different places around the slide. I am correcting for uniform illumination, using the product as directed, and sealing the coverslip with nail polish.

The nail polish may be the problem. The Kd value (calcium sensitivity) changes depending upon the dye's environment. Nail polish has solvents that can leech under the coverslip and cause variability. We recommend either going without a sealing or sealing with melted paraffin painted on the coverslip edges with a cotton-tipped applicator (paraffin is hydrophobic and has no solvents).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I need to label cells with Fluo-4, AM, for a calcium flux assay. How long after labeling will the dye be retained?

After loading dye into the cells, intracellular esterases remove the 'AM' moiety from the dye. When the 'AM' group is removed, the dye is able to bind calcium and fluoresce. Since the dye is not covalently bound to any cellular components, it may be actively effluxed from the cell. The rate of efflux is dependent upon the inherent properties of the cell, culture conditions and other factors. The dye may be retained for hours, days or even weeks or lost in a matter of minutes. The use of Probenecid (Cat. No. P36400) limits loss by active efflux.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (20)

Citations & References
Abstract
A light-dependent increase in free Ca2+ concentration in the salamander rod outer segment.
Authors:Matthews HR, Fain GL
Journal:J Physiol
PubMed ID:11306652
'1. The Ca(2+) indicator dye fluo-5F was excited by an argon ion laser to measure changes in free Ca(2+) concentration ([Ca2+]i) in the outer segments of isolated salamander rods rapidly exposed to a 0 Ca(2+), 0 Na(+) solution designed to minimise surface membrane Ca(2+) fluxes. Over 30-60 s of laser ... More
Catheter lock solutions influence staphylococcal biofilm formation on abiotic surfaces.
Authors:Shanks RM, Sargent JL, Martinez RM, Graber ML, O'Toole GA,
Journal:Nephrol Dial Transplant
PubMed ID:16627606
'BACKGROUND: Microbial biofilms form on central venous catheters and may be associated with systemic infections as well as decreased dialysis efficiency due to catheter thrombosis. The most widely used anticoagulant catheter lock solution in the US is sodium heparin. We have previously shown that sodium heparin in clinically relevant concentrations ... More
Dopaminergic regulation of dendritic calcium: fast multisite calcium imaging.
Authors:Zhou WL, Oikonomou KD, Short SM, Antic SD,
Journal:Methods Mol Biol
PubMed ID:23296782
'Optimal dopamine tone is required for the normal cortical function; however it is still unclear how cortical-dopamine-release affects information processing in individual cortical neurons. Thousands of glutamatergic inputs impinge onto elaborate dendritic trees of neocortical pyramidal neurons. In the process of ensuing synaptic integration (information processing), a variety of calcium ... More
The effect of light on outer segment calcium in salamander rods.
Authors:Matthews HR, Fain GL
Journal:J Physiol
PubMed ID:12949220
'Calcium acts as a second messenger in vertebrate rods, regulating the recovery phase of the light response and modulating sensitivity during light-adaptation. Since light not only decreases the outer segment calcium concentration ([Ca2+]i) by closing cyclic nucleotide-gated channels but can also increase [Ca2+]i by releasing Ca2+ from buffer sites or ... More
Imaging calcium entry sites and ribbon structures in two presynaptic cells.
Authors:Zenisek D, Davila V, Wan L, Almers W
Journal:J Neurosci
PubMed ID:12684438
'We investigated the location of calcium entry sites and synaptic ribbons in the type-Mb goldfish bipolar neuron and the bullfrog saccular hair cell. Cells were loaded with a fast calcium indicator (Fluo-3 or Fluo-5F) and an excess of a high-affinity but slow Ca buffer (EGTA). The cell surface was imaged ... More