FluoZin™-3, AM, cell permeant
Invitrogen™

FluoZin™-3, AM, cell permeant

Zn2+-selective indicator with a structure that resembles fluo-4.

Have Questions?
Catalog NumberQuantity
F24195100 μg
Catalog number F24195
Price (USD)
338.65
Online exclusive
360.00
Save 21.35
Each
Quantity:
100 μg
Price (USD)
338.65
Online exclusive
360.00
Save 21.35
Each

The FluoZin™-3 indicator is a Zn2+-selective indicator with a structure that resembles fluo-4. FluoZin™-3 exhibits high Zn2+-binding affinity that is unperturbed by Ca2+ concentrations up to at least 1 μM. In addition, FluoZin™-3 exhibits a >50-fold increase in fluorescence in response to saturating levels of Zn2+. Although imaging applications of FluoZin™-3 are predominant, cell-based microplate assays and flow cytometry protocols have also been developed. FluoZin™-3 indicator is available in a cell-permeant and a cell-impermeant form.

Fluorescent Zinc Indicators Specifications:

  • Label (Ex/Em): FluoZin™-3 (∼494/516 nm)
  • Kd (Zn2+) (in buffer): ∼15 nM
  • Lyophilized product may be dissolved in DMSO for use
  • Product is typically loaded into cells by adding the dissolved indicator to medium containing cells.

Find Fluorescent Indicators for Zinc and Other Metal Ions

We offer a number of fluorescent indicators to determine polyvalent cation concentrations inside cells, to follow metal ion transport through ion channels, to make measurements in environmental samples, and more. Review Fluorescent Indicators for Zn2+ and Other Metal Ions—Section 19.7 in the Molecular Probes™ Handbook for more information on these products.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeFluoZin-3
Emission Wavelength516 nm
Excitation Wavelength494 nm
Molecular Weight (g/mol)982.85 g/mol
Quantity100 μg
Shipping ConditionRoom Temperature
Emission Wavelength Range510 to 525 nm
Excitation Wavelength Range480 to 495 nm
For Use With (Application)Zinc Indicator
For Use With (Equipment)Fluorescence Microscope
Product LineFluoZin
Product TypeZinc Indicator
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

Why don't I see a significant change in signal for my live-cell fluorescent indicator dye?

Regardless of the type of live-cell indicator dye (e.g., calcium indicators, pH indicator, metal ion indicators), make sure there is no serum during the loading step, which can prematurely cleave dyes with AM esters and bind dyes non-specifically. Always optimize the dye concentration and staining time with a positive control before you run your test samples, to give the best signal-to-background. Always run a positive control with a buffer containing free ions of known concentration and an ionophore to open pores to those ions (for instance, for calcium indicators like Fluo-4 AM, this would include a buffer with added calcium combined with calcimycin, or for pH indicators, buffers of different pHs combined with nigericin). Reactive oxygen indicators, such as CellROX Green or H2DCFDA would require a cellular reactive oxygen species (ROS) stimulant as a positive control, such as menadione. Finally, make sure your imaging system has a sensitive detector. Plate readers, for instance, have much lower detector efficiency over background, compared to microscopy or flow cytometry.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (43)

Citations & References
Abstract
Demand for Zn2+ in acid-secreting gastric mucosa and its requirement for intracellular Ca2+.
Authors:Liu J, Kohler JE, Blass AL, Moncaster JA, Mocofanescu A, Marcus MA, Blakely EA, Bjornstad KA, Amarasiriwardena C, Casey N, Goldstein LE, Soybel DI,
Journal:PLoS One
PubMed ID:21698273
'Recent work has suggested that Zn(2+) plays a critical role in regulating acidity within the secretory compartments of isolated gastric glands. Here, we investigate the content, distribution and demand for Zn(2+) in gastric mucosa under baseline conditions and its regulation during secretory stimulation. Content and distribution of zinc were evaluated ... More
The amyloid precursor protein (APP) of Alzheimer disease and its paralog, APLP2, modulate the Cu/Zn-Nitric Oxide-catalyzed degradation of glypican-1 heparan sulfate in vivo.
Authors:Cappai R, Cheng F, Ciccotosto GD, Needham BE, Masters CL, Multhaup G, Fransson LA, Mani K
Journal:J Biol Chem
PubMed ID:15677459
'Processing of the recycling proteoglycan glypican-1 involves the release of its heparan sulfate chains by copper ion- and nitric oxide-catalyzed ascorbate-triggered autodegradation. The Alzheimer disease amyloid precursor protein (APP) and its paralogue, the amyloid precursor-like protein 2 (APLP2), contain copper ion-, zinc ion-, and heparan sulfate-binding domains. We have investigated ... More
Clioquinol and pyrithione activate TRPA1 by increasing intracellular Zn2+.
Authors:Andersson DA, Gentry C, Moss S, Bevan S,
Journal:Proc Natl Acad Sci U S A
PubMed ID:19416844
'The antifungal and amoebicidal drug clioquinol (CQ) was withdrawn from the market when it was linked to an epidemic of subacute myelo-optico-neuropathy (SMON). Clioquinol exerts its anti-parasitic actions by acting as a Cu/Zn chelator and ionophore. Here we show that local injections of CQ produce mechanical hyperalgesia and cold hypersensitivity ... More
Intracellular zinc fluctuations modulate protein tyrosine phosphatase activity in insulin/insulin-like growth factor-1 signaling.
Authors:Haase H, Maret W
Journal:Exp Cell Res
PubMed ID:14644152
'Zinc is an effector of insulin/IGF-1 signaling and has insulinomimetic effects, the molecular basis of which is not understood. The present study establishes the capacity of zinc to inhibit protein tyrosine phosphatases (PTPs) as a cause for these effects and, moreover, demonstrates modulation of the insulin response by changes in ... More
Interleukin-6 regulates the zinc transporter Zip14 in liver and contributes to the hypozincemia of the acute-phase response.
Authors:Liuzzi JP, Lichten LA, Rivera S, Blanchard RK, Aydemir TB, Knutson MD, Ganz T, Cousins RJ
Journal:Proc Natl Acad Sci U S A
PubMed ID:15863613
'Infection and inflammation produce systemic responses that include hypozincemia and hypoferremia. The latter involves regulation of the iron transporter ferroportin 1 by hepcidin. The mechanism of reduced plasma zinc is not known. Transcripts of the two zinc transporter gene families (ZnT and Zip) were screened for regulation in mouse liver ... More