FungaLight™ Yeast CFDA, AM/Propidium Iodide Vitality Kit - FAQs
View additional product information for FungaLight™ Yeast CFDA, AM/Propidium Iodide Vitality Kit - FAQs (F34953)
1 product FAQs found
Can I look at bacteria and yeast on a flow cytometer?
Yes, you can. We offer:
-Counting assays: Bacteria Counting Kit, for flow cytometry (Cat. No. B7277) or LIVE/DEAD BacLight Bacterial Viability and Counting Kit, for flow cytometry (Cat. No. L34856).
-Viability/vitality assays: LIVE/DEAD BacLight Bacterial Viability Kit (Cat. No. L13152).
-Membrane potential: BacLight Bacterial Membrane Potential Kit (Cat. No. B34950).
-Yeast viability/vitality assays: LIVE/DEAD FungaLight Yeast Viability Kit, for flow cytometry (Cat. No. L34952), FungaLight Yeast CFDA, AM/Propidium Iodide Vitality Kit (Cat. No. F34953)