Product Image
Thermo Scientific™

DyNAmo HS SYBR Green qPCR Kit

**********************Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
F410L500 Reactions
F410XL2500 Reactions
Catalog number F410L
Price (USD)
628.00
Each
No. of Reactions:
500 Reactions
Price (USD)
628.00
Each
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of DyNAmo HS SYBR Green qPCR Kit and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific DyNAmo HS SYBR Green qPCR Kit contains a modified Thermus brockianus DNA polymerase and all the other reagents needed for qPCR. Only template DNA and PCR primers need to be added by the user. The modified DNA polymerase in this kit incorporates a non-specific DNA-binding domain that lends physical stability to the polymerase-DNA complex.

ROX passive reference dye is also supplied with the kit in a separate tube for instruments that require ROX normalization.

Highlights

• Strong specificity: DyNAmo HS SYBR Green master mix minimizes the formation of unwanted primer-dimers and extension of non-specifically bound primers during reaction setup
• Highly sensitive detection: Optimized master mix allows reproducible detection of low-copy number templates
• Broad dynamic range: DyNAmo HS SYBR Green qPCR Kit detects as few as one copy of the target gene and achieves a dynamic range of up to eight orders of magnitude
• Convenient to use: 2X master mix, one tube only. ROX passive reference dye provided in a separate vial

Related Products
DyNAmo Flash Probe qPCR Kit
DyNAmo Flash SYBR Green qPCR Kit
DyNAmo HS SYBR Green qPCR Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, StepOne™ System, Stratagene Mx4000, Stratagene Mx3005P, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, Cepheid SmartCycler
FormatTube
Kit TypeDyNAmo HS SYBR Green qPCR Kit
No. of Reactions500 Reactions
PolymeraseTbr DNA polymerase
Product LineDyNAmo
Product TypeReal Time PCR SYBR Master Mix
Quantity500 reactions
Sample TypeDNA, dsDNA
Sufficient For500 Reactions of 20 μL Each, 200 Reactions of 50 μL Each
Volume5 x 1 mL
Concentration2X (qPCR Mix), 50X (Dye)
Detection MethodSYBR
For Use With (Application)Gene Expression
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Contains:
• 5 x1 mL DyNAmo HS SYBR Green qPCR Mix (2X)
• 250 μL 50X ROX dye, 25μM

Store at -20°C