BODIPY™ FL GDP (Guanosine 5'-Diphosphate, BODIPY™ FL 2'-(or-3')-O-(N-(2-Aminoethyl) Urethane), Bis (Triethylammonium) Salt)
The mixed-isomer analog of BODIPY® FL GDP is comprised of the BODIPY® FL fluorophore attached to the 2’ or 3’Read more
Have Questions?
Catalog Number
Quantity
G22360
also known as G-22360
100 μL
Catalog number G22360
also known as G-22360
Price (MXN)
-
Quantity:
100 μL
The mixed-isomer analog of BODIPY® FL GDP is comprised of the BODIPY® FL fluorophore attached to the 2’ or 3’ position of the ribose ring via an aminoethylcarbamoyl linker. BODIPY® FL exhibits spectral characteristics similar to fluorescein (FITC) or the Alexa Fluor® 488 dye.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeBODIPY™ FL
Product TypeFL GDP
Quantity100 μL
Shipping ConditionWet Ice
Concentration5 mM
Product LineBODIPY
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.
Citations & References (11)
Citations & References
Abstract
Capillary electrophoresis assay for G protein-coupled receptor-mediated GTPase activity.
'We describe a capillary electrophoresis (CE) assay to detect G protein-coupled receptor (GPCR)-stimulated G protein GTPase activity in cell membranes expressing alpha2A adrenoreceptor-Galphao1 wild-type (wt) or C351I mutant fusion proteins using a fluorescent, hydrolyzable GTP analogue. As no change in total fluorescence is observed by conversion of substrate to product, ... More
Role and timing of GTP binding and hydrolysis during EF-G-dependent tRNA translocation on the ribosome.
Authors:Wilden B, Savelsbergh A, Rodnina MV, Wintermeyer W
Journal:Proc Natl Acad Sci U S A
PubMed ID:16940356
The translocation of tRNA and mRNA through the ribosome is promoted by elongation factor G (EF-G), a GTPase that hydrolyzes GTP during the reaction. Recently, it was reported that, in contrast to previous observations, the affinity of EF-G was much weaker for GTP than for GDP and that ribosome-catalyzed GDP-GTP ... More
Probing the Ligand-Binding Pocket of BTN3A1.
Authors:
Journal:J Med Chem
PubMed ID:31268699
Protocol for structural and biochemical analyses of RhoA GTPase.
Authors:
Journal:STAR Protoc
PubMed ID:34036285
The small molecule ISRIB rescues the stability and activity of Vanishing White Matter Disease eIF2B mutant complexes.