This product will be discontinued on August 1, 2026. For routine DNA and RNA electrophoresis and imaging, we recommend the E-Gel™ Power Snap Lite Electrophoresis System, an updated platform optimized for use with 11- to 22-well precast E-Gel™ agarose gels.
E-Gel™ Power Snap Electrophoresis Device Starter Kit, SizeSelect II 2%
Invitrogen™
E-Gel™ Power Snap Electrophoresis Device Starter Kit, SizeSelect II 2%
Designed for fast and convenient E-Gel™ agarose gel electrophoresis, from DNA sample loading to gel image. Includes E-Gel™ SizeSelect™ II agarose gel, 2%.
Have Questions?
Catalog NumberQuantity
G8162ST1 Kit
Catalog number G8162ST
Price (ZAR)
-
Quantity:
1 Kit
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit with 2% E-Gel SizeSelect II agarose gels provides everything you need for faster and more effective DNA fragment library size selection in a next-generation sequencing (NGS) workflow.
The kit includes:
  • 10 E-Gel SizeSelect II agarose gels, 2% (Cat. No. G661012)
  • E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
  • E-Gel Sizing DNA Ladder (Cat. No. 10488100)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Kit
Shipping ConditionRoom Temperature
Voltage240 VAC
Max. Voltage48 VDC
TypeElectrophoresis System Starter Kit
Unit SizeEach
Contents & Storage
• 1 electrophoresis device
• 10 E-Gel SizeSelect II gels
• 1 vial E-Gel Sizing DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses
Store at room temperature.