E-Gel™ Power Snap Lite Electrophoresis System
E-Gel™ Power Snap Lite Electrophoresis System
E-Gel™ Power Snap Lite Electrophoresis System
Invitrogen™

E-Gel™ Power Snap Lite Electrophoresis System

The Invitrogen™ E-Gel™ Power Snap Lite integrated system and its individual components enable a fast, convenient workflow for loading, running, and analyzing DNA samples on precast E-Gel™ agarose gels, from sample loading to gel imaging. Designed for routine horizontal electrophoresis, it supports quick verification and workflow checks with agarose gels for up to 22 samples per run.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
G8700Horizontal Gel Electrophoresis System
G8502Horizontal Agarose Electrophoresis Unit
G8600E-Gel Camera
Catalog number G8700
Price (USD)
2,717.00
Special offer
Online exclusive
Ends: 04-Dec-2026
4,180.00
Save 1,463.00
Each
Type:
Horizontal Gel Electrophoresis System
Price (USD)
2,717.00
Special offer
Online exclusive
Ends: 04-Dec-2026
4,180.00
Save 1,463.00
Each

The Invitrogen™ E-Gel™ Power Snap Lite system, consisting of the E-Gel™ Power Snap Lite Electrophoresis Device* and E-Gel™ Power Snap Lite Camera*, enables fast, convenient E-Gel™ agarose gel electrophoresis for up to 22 samples per run, from sample loading to gel imaging. Using dry, precast E-Gel agarose gel technology, DNA samples can be run in as little as 10 minutes while separation is monitored in real time. This benchtop system integrates DNA separation and gel imaging into a single workflow.

Features of the E-Gel Power Snap Lite Electrophoresis System include:

  • Streamlined electrophoresis and image capture in one integrated system
  • DNA and RNA separation in as little as 10 minutes with dry, precast E-Gel agarose gels
  • Touchscreen interface with preset protocols for fast, consistent runs
  • Real-time viewing and instant capture E-Gel agarose gel images
  • Recycled plastic components to support your sustainability goals

Small, fast, and powerful

The all-in-one E-Gel Power Snap Lite Electrophoresis System supports DNA gel electrophoresis, gel visualization and imaging in a compact, lightweight benchtop instrument. The system includes an integrated power supply, blue-light transilluminator, and amber filter, enabling gel separation and real-time sample tracking on E-Gel agarose gels pre-stained with SYBR Safe or SYBR Gold II DNA stains. It also comes with preprogrammed protocols for each available E-Gel agarose gel type.

Dry, precast E-Gel agarose gels for added convenience

With dry, precast, pre-stained E-Gel agarose gels, DNA or RNA samples can be run in as little as 10 minutes. This technology helps save time and simplify workflows by eliminating the need for gel preparation and staining. E-Gel agarose gels are available in multiple gel percentages and with a choice of two DNA stains.

The E-Gel Power Snap Lite Electrophoresis System is compatible with low-throughput E-Gel precast agarose gels, including E-Gel EX, E-Gel SYBR Safe, E-Gel CloneWell II, and E-Gel SizeSelect II gels.

Instant image capture

The included E-Gel Power Snap Lite Camera* makes it easy to control gel runs and capture high-resolution images as you work. The camera docks directly to the E-Gel Power Snap Lite Electrophoresis Device and requires no external power supply or desktop computer connection, allowing you to manage the entire gel run workflow right at the bench.

Designed for convenience

The E-Gel Power Snap Lite Camera touchscreen and onboard software allow an intuitive interface for fast image capture and editing. Images can be stored in the camera's internal memory for months of use or transferred easily to an external USB storage device. The E-Gel Power Snap Lite Camera includes an automatic gel capture workflow that enables multiple images to be captured at defined time intervals while you are away from the bench. When you return, your images are available in the internal image gallery.

An ideal choice for cloning workflows

The blue-light transilluminator in the E-Gel Power Snap Lite Electrophoresis Device helps maximize user safety and can improve cloning efficiency compared with ethidium bromide/UV when used with E-Gel CloneWell II agarose gels.

Product warranty

The Invitrogen™ E-Gel™ Power Snap Lite system and its individual components include a 12-month warranty.

*The E-Gel Power Snap Lite Device is compatible only with the E-Gel Power Snap Lite Camera. Neither product is compatible with other cameras in the E-Gel Power Snap family. Both products are intended exclusively for E-Gel agarose gel electrophoresis and imaging.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferNo Buffer Needed
Detection MethodFluorescence in Blue Light
Dimensions (L x W x H)260 x 130 x 200 mm
DisplayTouchscreen LCD Display
Frequency50/60 Hz
Gel Percentage0.8 to 4%
Gel TypeE-Gel Agarose Gel
IncludesElectrophoresis Device, Camera, Power Cord, Viewing Glasses
Label or DyeSYBR Safe, SYBR Gold II
Max. Current1.87 A
Mode of SeparationElectrophoresis
Quantity1 Ea.
Sample TypeDNA, RNA
Shipping ConditionRoom Temperature
Timer0 to 99 min. or continuous
Warranty12 Month
Weight (Metric)2.7 kg
CapacityUp to 22 Samples in E-Gel Agarose Gel
For Use With (Application)Gel Electrophoresis, Nucleic Acid Analysis, Demonstrating Horizontal Gel Electrophoresis to Students
For Use With (Equipment)No Dependencies
Gel CompatibilityE-Gel Agarose Gels with SYBR Dyes
Gel Size10 x 8 cm
Max. Voltage48 VDC
Product LineE-Gel
Running Time5 to 30 min.
TypeHorizontal Gel Electrophoresis System
Unit SizeEach