Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
GAU 010-04-02 detects Complement factor D in human samples.
GAU 010-04-02 has been successfully used in Western blot and as a capture antibody in sandwich ELISA with GAU 008-01-02 as detection antibody.
Complement factor D is a serine protease of the alternative pathway of complement activation. The protein is a single chain polypeptide with a molecular mass of 25 kDa. Factor D cleaves factor B bound to C3b, generating the alternative pathway C3 convertase C3bBb and releasing the Ba fragment.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adipsin; C3 convertase activator; Complement factor D; complement factor D preproprotein; D component of complement (adipsin); DF; EC 3.4.21.46; Properdin factor D; unnamed protein product
Gene Aliases: ADIPSIN; ADN; CFD; DF; PFD
UniProt ID: (Human) P00746
Entrez Gene ID: (Human) 1675
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support