Search
Search
Fabgennix
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
GGCX (gamma-glutamyl carboxylase), also known as GC or VKCFD1 (Vitamin K-dependent gamma-carboxylase), is a 758 amino acid multi-pass membrane protein. Localized to the membrane of the endoplasmic reticulum, GGCX functions to mediate the vitamin K-dependent carboxylation of glutamate residues on target proteins, thereby producing calcium binding gamma-carboxyglutamate (Gla) residues on these proteins and simultaneously converting vitamin K to vitamin K epoxide. GGCX exists as a monomer and, via its ability to modify glutamate residues, it accomplishes the post-translational changes that are necessary for the activity of all vitamin K-dependent proteins (such as blood coagulation and bone matrix proteins). Defects in the gene encoding GGCX are the cause of combined deficiency of vitamin K-dependent clotting factors 1 (VKCFD1) and PXE-like disorder with multiple coagulation factor deficiency, both of which are characterized by abnormal skin, blood or bone function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ26629; Gamma-glutamyl carboxylase; Peptidyl-glutamate 4-carboxylase; unnamed protein product; Vitamin K gamma glutamyl carboxylase; Vitamin K-dependent gamma-carboxylase; VKGC
Gene Aliases: GC; GGCX; VKCFD1
UniProt ID: (Human) P38435, (Rat) O88496, (Mouse) Q9QYC7
Entrez Gene ID: (Human) 2677, (Rat) 81716, (Mouse) 56316
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support