Human MATE1 (G) SLC Transporter Cells - FAQs
Human MATE1 (G) SLC Transporter Cells - FAQs
View additional product information for Human MATE1 (G) SLC Transporter Cells - FAQs (GM1114G)
6 product FAQs found
What are TRANSiPORT human solute carrier (SLC) transporter cells?
TRANSiPORT human SLC transporter cells are HEK293 cells that transiently overexpress a solute carrier (SLC) transporter protein. Solute carriers are one of two superfamilies of membrane transporters that can affect pharmacokinetics and drug exposure by governing the transport of solutes into and out of cells. Membrane transporters are found in hepatic cells, kidney proximal tubule cells, intestinal cells, and brain cells.
Find additional tips, troubleshooting help, and resources within our Drug Discovery & Development Support Center.
Is there a protocol available for the TRANSiPORT solute carrier (SLC) transporter cells?
Yes, the protocol is available here (https://www.genomembrane.com/information).
What should I use as a control with TRANSiPORT solute carrier (SLC) transporter cells?
Mock HEK293 cells (Cat. No. GM1001) that do not overexpress the SLC transporter are available for use as a negative experimental control. Some compounds may demonstrate high background levels of SLC transport due to the presence of endogenous transporters or non-specific binding.
With TRANSiPORT solute carrier (SLC) transporter cells, what should my choice of measurement be for SLC transporter activity?
Cell-based assays can be performed using radioisotope-labeled compounds, fluorescence-labeled compounds, or non-labeled compounds. The amount of substrate transported into the cells can be measured directly using a liquid scintillation counter, fluorescence plate reader, or LC-MS/MS, thereby allowing direct evaluation of SLC transporter activity.
How many cells can I expect in one vial of TRANSiPORT solute carrier (SLC) transporter cells?
One vial contains 8-12 million cells with high viability, designed to seed one 24-well plate for single use.
What are the shipping and storage conditions for TRANSiPORT solute carrier (SLC) transporter cells?
Domestic shipments are on dry ice and shipped overnight. We ask our customers to store the product under liquid nitrogen after they receive it. International shipments are under liquid nitrogen conditions.