Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: KAIARLIEFY KHESCGQCTP CREGVDWMNK VMARFVRGDA RPAEIDSLWE ISKQIEGHTI CALGDGAAWP VQGLIRHFRP ELEERMQRFA QQHQARQAAS
NDUFV1 is the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.The NDUFV1 gene encodes the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 51 kda flavoprotein; CI-51kD; complex I 51 kda subunit; complex I 51kDa subunit; complex I, mitochondrial respiratory chain; Complex I-51kD; mitochondrial NADH dehydrogenase ubiquinone flavoprotein 1; mitochondrial NADH:ubiquinone oxidoreductase 51 kda subunit; NADH dehydrogenase (ubiquinone) flavoprotein 1, 51kDa; NADH dehydrogenase [ubiquinone] flavoprotein 1, mitochondrial; NADH dehydrogenase flavoprotein 1; NADH-ubiquinone oxidoreductase 51 kDa subunit; NDUFV1; unnamed protein product
Gene Aliases: CI-51K; CI51KD; MC1DN4; NDUFV1; UQOR1
UniProt ID: (Human) P49821
Entrez Gene ID: (Human) 4723, (Rat) 293655
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support