Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • ARGBP2 Antibodies

          Abnova

          SORBS2 Polyclonal Antibody, MaxPab™

          View all (7) ARGBP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SORBS2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SORBS2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SORBS2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SORBS2 Antibody (H00008470-B01P) in WB

          Western Blot analysis of SORBS2 expression in transfected 293T cell line by SORBS2 MaxPab polyclonal antibody. Lane 1: SORBS2 transfected lysate(121 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SORBS2 Antibody in Western Blot (WB)
          SORBS2 Polyclonal Antibody, MaxPab™

          Product Details

          H00008470-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SORBS2 (NP_066547.1, 1 a.a. approximately 1100 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSYYQRPFSP SAYSLPASLN SSIVMQHGTS LDSTDTYPQH AQSLDGTTSS SIPLYRSSEE EKRVTVIKAP HYPGIGPVDE SGIPTAIRTT VDRPKDWYKT MFKQIHMVHK PDDDTDMYNT PYTYNAGLYN PPYSAQSHPA AKTQTYRPLS KSHSDNSPNA FKDASSPVPP PHVPPPVPPL RPRDRSSTEK HDWDPPDRKV DTRKFRSEPR SIFEYEPGKS SILQHERPAS LYQSSIDRSL ERPMSSASMA SDFRKRRKSE PAVGPPRGLG DQSASRTSPG RVDLPGSSTT LTKSFTSSSP SSPSRAKGGD DSKICPSLCS YSGLNGNPSS ELDYCSTYRQ HLDVPRDSPR AISFKNGWQM ARQNAEIWSS TEETVSPKIK SRSCDDLLND DCDSFPDPKV KSESMGSLLC EEDSKESCPM AWGSPYVPEV RSNGRSRIRH RSARNAPGFL KMYKKMHRIN RKDLMNSEVI CSVKSRILQY ESEQQHKDLL RAWSQCSTEE VPRDMVPTRI SEFEKLIQKS KSMPNLGDDM LSPVTLEPPQ NGLCPKRRFS IEYLLEEENQ SGPPARGRRG CQSNALVPIH IEVTSDEQPR AHVEFSDSDQ DGVVSDHSDY IHLEGSSFCS ESDFDHFSFT SSESFYGSSH HHHHHHHHHH RHLISSCKGR CPASYTRFTT MLKHERARHE NTEEPRRQEM DPGLSKLAFL VSPVPFRRKK NSAPKKQTEK AKCKASVFEA LDSALKDICD QIKAEKKRGS LPDNSILHRL ISELLPDVPE RNSSLRALRR SPLHQPLHPL PPDGAIHCPP YQNDCGRMPR SASFQDVDTA NSSCHHQDRG GALQDRESPR SYSSTLTDMG RSAPRERRGT PEKEKLPAKA VYDFKAQTSK ELSFKKGDTV YILRKIDQNW YEGEHHGRVG IFPISYVEKL TPPEKAQPAR PPPPAQPGEI GEAIAKYNFN ADTNVELSLR KGDRVILLKR VDQNWYEGKI PGTNRQGIFP VSYVEVVKKN TKGAEDYPDP PIPHSYSSDR IHSLSSNKPQ RPVFTHENIQ GGGEPFQALY NYTPRNEDEL ELRESDVIDV MEKCDDGWFV GTSRRTKFFG TFPGNYVKRL

          Target Information

          Arg and c-Abl represent the mammalian members of the Abelson family of non-receptor protein-tyrosine kinases. They interact with the Arg/Abl binding proteins via the SH3 domains present in the carboxy end of the latter group of proteins. ARGBP2 is the sorbin and SH3 domain containing 2 protein. It has three C-terminal SH3 domains and an N-terminal sorbin homology(SoHo) domain that interacts with lipid raft proteins. The subcellular localization of this protein in epithelial and cardiac muscle cells suggests that it functions as an adapter protein to assemble signaling complexes in stress fibers, and that it is a potential link between Abl family kinases and the actin cytoskeleton.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Arg binding protein 2; Arg-binding protein 2; Arg/Abl-interacting protein 2; ArgBP2; Sorbin; Sorbin and SH3 domain-containing protein 2; unnamed protein product

          View more View less

          Gene Aliases: ARGBP2; KIAA0777; PRO0618; SORBS2

          View more View less

          UniProt ID: (Human) O94875

          View more View less

          Entrez Gene ID: (Human) 8470

          View more View less

          Function(s)
          RNA binding structural constituent of cytoskeleton protein binding cytoskeletal adaptor activity structural constituent of muscle identical protein binding
          Process(es)
          cytoskeleton organization Notch signaling pathway cell growth involved in cardiac muscle cell development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline