Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SRGAP1 Antibodies

          Abnova

          SRGAP1 Polyclonal Antibody, MaxPab™

          View all (8) SRGAP1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SRGAP1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          SRGAP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SRGAP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SRGAP1 Antibody (H00057522-B01P) in ICC/IF

          Immunofluorescence of purified MaxPab antibody to SRGAP1 on HeLa cell. Antibody concentration is 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SRGAP1 Antibody in Immunocytochemistry (ICC/IF)
          SRGAP1 Antibody in Western Blot (WB)
          SRGAP1 Polyclonal Antibody, MaxPab™

          Product Details

          H00057522-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SRGAP1 (NP_065813.1, 1 a.a. approximately 1085 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MSTPSRFKKD KEIIAEYESQ VKEIRAQLVE QQKCLEQQTE MRVQLLQDLQ DFFRKKAEIE TEYSRNLEKL AERFMAKTRS TKDHQQYKKD QNLLSPVNCW YLLLNQVRRE SKDHATLSDI YLNNVIMRFM QISEDSTRMF KKSKEIAFQL HEDLMKVLNE LYTVMKTYHM YHAESISAES KLKEAEKQEE KQIGRSGDPV FHIRLEERHQ RRSSVKKIEK MKEKRQAKYS ENKLKSIKAR NEYLLTLEAT NASVFKYYIH DLSDLIDCCD LGYHASLNRA LRTYLSAEYN LETSRHEGLD IIENAVDNLE PRSDKQRFME MYPAAFCPPM KFEFQSHMGD EVCQVSAQQP VQAELMLRYQ QLQSRLATLK IENEEVKKTT EATLQTIQDM VTIEDYDVSE CFQHSRSTES VKSTVSETYL SKPSIAKRRA NQQETEQFYF MKLREYLEGS NLITKLQAKH DLLQRTLGEG HRAEYMTTRP PNVPPKPQKH RKSRPRSQYN TKLFNGDLET FVKDSGQVIP LIVESCIRFI NLYGLQHQGI FRVSGSQVEV NDIKNSFERG ENPLADDQSN HDINSVAGVL KLYFRGLENP LFPKERFNDL ISCIRIDNLY ERALHIRKLL LTLPRSVLIV MRYLFAFLNH LSQYSDENMM DPYNLAICFG PTLMPVPEIQ DQVSCQAHVN EIIKTIIIHH ETIFPDAKEL DGPVYEKCMA GDDYCDSPYS EHGTLEEVDQ DAGTEPHTSE DECEPIEAIA KFDYVGRSAR ELSFKKGASL LLYHRASEDW WEGRHNGIDG LVPHQYIVVQ DMDDTFSDTL SQKADSEASS GPVTEDKSSS KDMNSPTDRH PDGYLARQRK RGEPPPPVRR PGRTSDGHCP LHPPHALSNS SVDLGSPSLA SHPRGLLQNR GLNNDSPERR RRPGHGSLTN ISRHDSLKKI DSPPIRRSTS SGQYTGFNDH KPLDPETIAQ DIEETMNTAL NELRELERQS TAKHAPDVVL DTLEQVKNSP TPATSTESLS PLHNVALRSS EPQIRRSTSS SSDTMSTFKP MVAPRMGVQL KPPALRPKPA VLPKTNPTIG PAPPPQGPTD KSCTM

          Target Information

          The protein encoded by this gene is a GTPase activator, working with the GTPase CDC42 to negatively regulate neuronal migration. The encoded protein interacts with the transmembrane receptor ROBO1 to inactivate CDC42.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Rho GTPase-activating protein 13; SLIT-ROBO Rho GTPase-activating protein 1; srGAP1

          View more View less

          Gene Aliases: ARHGAP13; KIAA1304; NMTC2; SRGAP1

          View more View less

          UniProt ID: (Human) Q7Z6B7

          View more View less

          Entrez Gene ID: (Human) 57522

          View more View less

          Function(s)
          GTPase activator activity protein binding small GTPase binding
          Process(es)
          signal transduction Rho protein signal transduction nervous system development cell migration negative regulation of cell migration regulation of synapse assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline