Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • MMRN2 Antibodies

          Abnova

          MMRN2 Polyclonal Antibody, MaxPab™

          View all (3) MMRN2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MMRN2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          MMRN2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MMRN2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MMRN2 Antibody (H00079812-B01P) in WB

          Western Blot analysis of MMRN2 expression in transfected 293T cell line by MMRN2 MaxPab polyclonal antibody. Lane 1: MMRN2 transfected lysate(104.39 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MMRN2 Antibody in Western Blot (WB)

          Product Details

          H00079812-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MMRN2 (AAH94744.1, 1 a.a. approximately 949 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MILSLLFSLG GPLGWGLLGA WAQASSTSLS DLQSSRTPGV WKAEAEDTGK DPVGRNWCPY PMSKLVTLLA LCKTEKFLIH SQQPCPQGAP DCQKVKVMYR MAHKPVYQVK QKVLTSLAWR CCPGYTGPNC EHHDSMAIPE PADPGDSHQE PQDGPVSFKP GHLAAVINEV EVQQEQQEHL LGDLQNDVHR VADSLPGLWK ALPGNLTAAV MEANQTGHEF PDRSLEQVLL PHVDTFLQVH FSPIWRSFNQ SLHSLTQAIR NLSLDVEANR QAISRVQDSA VARADFQELG AKFEAKVQEN TQRVGQLRQD VEDRLHAQHF TLHRSISELQ ADVDTKLKRL HKAQEAPGTN GSLVLATPGA GARPEPDSLQ ARLGQLQRNL SELHMTTARR EEELQYTLED MRATLTRHVD EIKELYSESD ETFDQISKVE RQVEELQVNH TALRELRVIL MEKSLIMEEN KEEVERQLLE LNLTLQHLQG GHADLIKYVK DCNCQKLYLD LDVIREGQRD ATRALEETQV SLDERRQLDG SSLQALQNAV DAVSLAVDAH KAEGERARAA TSRLRSQVQA LDDEVGALKA AAAEARHEVR QLHSAFAALL EDALRHEAVL AALFGEEVLE EMSEQTPGPL PLSYEQIRVA LQDAASGLQE QALGWDELAA RVTALEQASE PPRPAEHLEP SHDAGREEAA TTALAGLARE LQSLSNDVKN VGRCCEAEAG AGAASLNASL DGLHNALFAT QRSLEQHQRL FHSLFGNFQG LMEANVSLDL GKLQTMLSRK GKKQQKDLEA PRKRDKKEAE PLVDIRVTGP VPGALGAALW EAGSPVAFYA SFSEGTAALQ TVKFNTTYIN IGSSYFPEHG YFRAPERGVY LFAVSVEFGP GPGTGQLVFG GHHRTPVCTT GQGSGSTATV FAMAELQKGE RVWFELTQGS ITKRSLSGTA FGGFLMFKT

          Target Information

          Inhibits endothelial cells motility and acts as a negative regulator of angiogenesis; it downregulates KDR activation by binding VEGFA.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Elastin microfibril interface located protein 3; Elastin microfibril interfacer 3; EMILIN-3; EMILIN-like protein EndoGlyx-1; EndoGlyx-1 p125/p140 subunit; Multimerin-2; unnamed protein product

          View more View less

          Gene Aliases: EMILIN-3; EMILIN3; ENDOGLYX-1; MMRN2

          View more View less

          UniProt ID: (Human) Q9H8L6

          View more View less

          Entrez Gene ID: (Human) 79812

          View more View less

          Function(s)
          extracellular matrix structural constituent protein binding receptor agonist activity
          Process(es)
          angiogenesis cell migration involved in sprouting angiogenesis cell adhesion signal transduction negative regulation of cell proliferation negative regulation of cell migration negative regulation of vascular endothelial growth factor receptor signaling pathway negative regulation of cell migration involved in sprouting angiogenesis positive regulation of defense response to bacterium negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis positive regulation of epithelial tube formation positive regulation of morphogenesis of an epithelium
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline