Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: FKSSRTEPHC IIINLSTWII WCYECDEKLS THCNKKVLAQ IVDFLQKHAS KTQTSAFSRI MKLCEEKCET DEIQKGGKCR NLSVRGITNL G
The protein encoded by this gene is a deubiquitylase that binds ERCC1, the catalytic subunit of the XPF-ERCC1 DNA repair endonuclease. This endonuclease is a critical regulator of DNA repair processes, and the deubiquitylase activity of the encoded protein is important for maintaining the DNA repair ability of XPF-ERCC1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 45; granule cell antiserum positive 7; Ubiquitin carboxyl-terminal hydrolase 45; ubiquitin specific petidase 45; ubiquitin specific protease 45; Ubiquitin thioesterase 45; ubiquitin thiolesterase 45; Ubiquitin-specific-processing protease 45
Gene Aliases: 3110003C05Rik; 4930550B20Rik; AI843191; Gcap7; LCA19; USP45
UniProt ID: (Human) Q70EL2, (Mouse) Q8K387
Entrez Gene ID: (Human) 85015, (Rat) 313098, (Mouse) 77593
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support