Can the Hypoxia Green Reagent for Flow Cytometry be used in cells expressing fluorescent proteins?
Yes, as long as the fluorescent protein does not emit in the green range (510–535 nm). For cells that do not express a fluorescent protein, we recommend viewing unstained cells under the FITC channel to examine autofluorescence. For cells expressing a fluorescent protein, we recommend analyzing unstained cell samples to determine the extent the fluorescent protein emission may overlap in the FITC channel.