HCS LipidTOX™ Phospholipidosis and Steatosis Detection Kit, for high-content screening, for cellular imaging - FAQs
HCS LipidTOX™ Phospholipidosis and Steatosis Detection Kit, for high-content screening, for cellular imaging - FAQs
View additional product information for HCS LipidTOX™ Phospholipidosis and Steatosis Detection Kit, for high-content screening, for cellular imaging - FAQs (H34158)
1 product FAQs found
I'm seeing aggregates in my thawed LipidTox stain solutions. Is this okay?
After thawing LipidTOX Red phospholipidosis detection reagent, some minute aggregates might be observed in the solution. They usually will disappear if the vial is incubated in a 37 degrees C water bath for 5 minutes. These aggregates do not affect the performance of the assay.
Any aggregates that remain after the stain is diluted in media are removed by 0.2 µm filtration before the labeling solution is added to the cells, as recommended in the protocol.