For a final staining solution, we can add TMRM directly into cell growth medium. Can this media contain serum?
Yes, you may add TMRM to media with serum. Serum components may bind dye non-specifically so you may need a higher final working concentration for staining samples in media containing serum relative to staining the same cells in a non-serum media or buffer.