Watson™ LIMS™ Software - FAQs
Watson™ LIMS™ Software - FAQs
View additional product information for Watson™ LIMS™ Software - FAQs (INF-21000)
29 product FAQs found
How do you perform and review a Standard Regression Analysis for PK studies and an Immune Response Analysis for ADA in Watson LIMS Software after importing raw results?
To perform and review Standard Regression Analysis for PK Studies and an Immune Response Analysis for ADA we recommend the following:
PK Studies:
Import raw results.
Perform Standard Regression Analysis to calculate the calibration curve and regression parameters.
Use a Run Acceptance Template to verify run acceptance criteria.
Review the Curve, Standards, and QCs. Flag ALQ and BLQ samples, and tag samples for re-assay if needed.
Save the form.
A second user reviews and accepts the results.
ADA Studies:
Follow a similar process as PK studies.
Perform Immune Response Analysis.
The first operator assesses and saves the form.
A second operator reviews and accepts the results.
Does Watson LIMS Software manage multiple aliquots of the same sample?
Watson LIMS Software manages multiple aliquots, called splits, of the same sample. This can be done both manually and by importing information through spreadsheet files.
Which file formats are available for results export (e.g .xml, .xls, .csv) in Watson LIMS Software?
You can use the Export function to export all data in the Explorer to one of the following formats: Excel, CSV, Rich Text Format, PDF and single web page file.
Can Watson LIMS Software do a check sum of the content of the export file?
For files exported by Watson LIMS Software, the system cannot add a checksum.
For files exported by another system to be imported by Watson LIMS, the ability of the system to add a checksum depends.
- Watson LIMS supports, through Integration Manager, the SOFT-interface (Secure Open-standard File Transfer) which works with Digital Signatures, which is more secure than a checksum. Currently, only a limited number of suppliers support this format. For build-in interfaces, no file validation is performed.
- For other interfaces implemented via Integration Manager a file verification is not standard, but can be added; this requires further input and collaboration from the instrument vendor.
Can Watson LIMS Software use LDAP / Single Sign On as authentication mechanism?
Watson LIMS Software can use LDAP and SSO for authentication.
How does Watson LIMS ensure accurate, complete, reliable, and compliant records and data throughout the data life cycle?
Watson LIMS Software is able to ensure accurate, complete, reliable, and compliant records and data throughout the data life cycle.
Can Watson LIMS Software provide the ability to receive and store samples including location and any further movement?
Watson LIMS Software is able to receive and store samples. These include locations and any further movement.
Are records protected against accidental modification or deletion in Watson LIMS Software?
Data is stored in a secured Oracle database.
Are there are any additional costs for “non production” environments in Watson LIMS Software?
There’s no additional cost for two additional non-productive systems (like DEV and VAL). These systems will only get a limited number of licenses (10% of the total). During the project phase a full set of licenses will be granted. If deployment tools are required (hosting fees, remoteApp, and licenses for Windows / Oracle, etc…) these will be charged separately.
Can you add comments to a sample in Watson LIMS Software at any stage of the process?
Watson LIMS Software allows you to apply comments to samples in the Worklist > All Samples Tab either by selecting from the drop-down list or by entering free text. This comment can be for any reason you choose.
Are Plate IDs / Assay IDs generated by Watson LIMS Software?
All Project ID’s/Plate IDs/Assay IDs that are generated by the system are unique.
What are the sample conditions available within Watson LIMS Software, and how can additional sample conditions be added to manage samples more accurately?
Sample Conditions within Watson LIMS Software includes Not Received, OK, and In Run. The condition changes automatically as you process. It is possible to add other sample conditions through the Watson Libraries, allowing you to manage your samples more accurately.
How can I export authorized results in Watson LIMS Software, and what does the Custom Results Export feature offer?
Authorized results can be exported through the Custom Results Export Feature. Custom Results Export produces a file of sample results with a customizable variety of information associated with each result.
In addition to the direct interface with Analyst, which systems already have an interface with Watson LIMS Software (MSD, Softmax etc)?
Watson LIMS Software has dozens of built-in file-based interfaces that can be used (Softmax Pro, MSD {single analyte}, Tecan, Sciex, Masslynx, etc.)
Besides the digital interface for Sciex Analyst, various so-called “gateways” are available for direct interfaces: LCQuan, XCalibur, Thermo Scientific TraceFinder Software, Thermo Scientific Chromeleon Chromatography Data System (CDS) Software, Empower.
Integration Manager provides bi-directional interfaces between LIMS, CDS, instruments and enterprise systems to allow automated data transfers from lab instruments into Watson LIMS Software.
Waters UNIFI is connected to Integration Manager in a file-less manner.
Thermo Fisher Scientific supports the SOFT-interface (Secure Open-standard File Transfer), which is a digitally signed (hence secured), standardized xml-file for interfacing between LIMS and instrument software.
For all other instruments a file-based transfer is possible using Integration Manager. This has been done for numerous instruments (MSD, Softmax Pro, Tecan, Simoa, Ella, Luminex, MassHunter, etc..)
How do you select samples for Incurred Sample Repeats (ISR)? Is Watson LIMS Software able to automatize the selection or to suggest some samples based on rules? And the minimum number of samples to be re-analysed?
Incurred Sample Repeats (ISR) sample selection can be randomly selected based on the results, whereby ALQ and/or BLQ results can be filtered out prior to the random selection. However, according to industry guidelines [ICH-M10] the minimum number of ISR is a dynamic rule:
- 10% of first 1,000 samples; 5% of all samples above 1,000
- randomly selected subjects
- adequate coverage of the concentration profile is important.
This pseudo-randomization makes it difficult to build into the system and most customers tag samples for ISR manually, so that they can be found more easily when selecting samples for a next run.
Does Watson LIMS Software provide the ability to import samples from an import file?
Watson LIMS Software is able to import samples from an import file. This can be done by the use of an excel file, for example.
Are backup, archiving, and restore operations tracked and documented (e.g., audit trail captured, logged) within Watson LIMS Software and available for review?
Watson LIMS Software allows for backup, archiving, and restore operations to be tracked and documented within the audit trail.
Does Watson LIMS Software ensure that GxP-related actions are performed in the correct sequence of business process steps and prevent non-permitted sequencing of steps and events?
Watson LIMS Software ensures that GxP related actions are performed in the correct sequence.
Can Watson LIMS Software generate assay reports that are printable and suitable for documenting assay procedures in the laboratory?
Assay reports within Watson LIMS Software that are generated can be saved and printed.
Can I add additional information during assay planning in Watson LIMS Software?
Within Watson LIMS Software, additional information can be added including Assay Type, Assay Date, Biological Matrix, Species, Analyte, Instrument type/interface, Operator and more.
Could you describe the study design within Watson LIMS Software?
Within Watson LIMS Software, flexible study designs are defined using a built-in protocol builder, using template-based study protocol import, and import using an API. This allows for the import of custom barcodes to keep track of protocol amendments and for Sample Design Reconciliation.
Which kind of actions are available in Watson LIMS Software once the study is completed /on-hold/in stand-by?
There are many different study actions available depending on the status of a study. Some of these include Archive Study, Assign Security, Configuration, Delete/Edit Study, Import/Export, Reassign Study, Rename Study, Study Analytes, Lock/Unlock Study, Electronic Signatures, Study Audit Report, Study Level Retriever Reports and Document Management System.
Once the Study is completed, it can be locked. Upon locking a separate (configurable) security role becomes active and will limit most actions to a Study – but will typically allow access to all reports. It’s also possible to revoke access to the Study for all users, except the archivist.
Does Watson LIMS Software provide the ability to create and open Calibration, QC, and Test Batches with or without spikes?
Watson LIMS Software is able to create and open Calibration, QC and Test Batches with or without spikes.
Does Watson LIMS Software provide the ability to log the reason for changes as part of the audit trail for designated actions?
Watson LIMS Software allows the reason for change to be logged. Also, in the use of electronic signatures, a reason for carrying out a change is presented as part of the audit trail and to keep track of any changes made.
How does Watson LIMS Software manage stability samples and what conditions are assessed for these samples?
Stability samples can be managed if they are defined as stability samples. Stability samples are samples that are maintained under strict conditions to assess the effect of environmental parameters such as temperature, humidity or freeze/thaw cycles. They can be processed in the same way as study samples and can be used in all types of analytical runs.
Is it possible to sort the samples in Watson LIMS Software during the creation of a worklist (for example sort by Day and timepoint) and during result review?
Samples are able to be sorted by multiple fields including day and time during both creation of a worklist and result review.
Can Watson LIMS Software use barcode identifiers (including 2D codes) for error-free identification of samples and materials?
Watson LIMS Software is able to use barcode identifier (including 2D codes and QR codes) for sample identification.
Can Watson LIMS Software be configured to achieve the required performance, allowing the user to create multi-analyte sequences in 96 and 384 well plates?
Watson LIMS Software is able to create multi analyte sequences in 96 and 384 wells plates.
Can you provide more details on QC batch creation and its use in worklist creation?
Yes, QCs are known as “Analytical Samples” and can be created in a batch. Labels can be generated and (optionally) QC-samples are tracked in the Sample Management area of Watson LIMS Software. As such QC-samples can be ‘received’, transferred and tracked for F/T-parameters and used in Stability Reports and other Method Validation Reports. It can be configured that after single use the QC-sample will be “consumed”.