LTQ XL™ Linear Ion Trap Mass Spectrometer
Thermo Scientific™
LTQ XL™ Linear Ion Trap Mass Spectrometer
This high-performance linear ion trap delivers excellent sensitivity and rich structural information.
Have Questions?
Catalog NumberType
IQLAAEGAAVFACZMAIKLTQ XL Linear Ion Trap MS
Catalog number IQLAAEGAAVFACZMAIK
Price (MXN)
-
Type:
LTQ XL Linear Ion Trap MS
Obtain high sensitivity full scan MS along with in-depth MSn (CRM) capabilities with the Thermo Scientific™ LTQ XL™ linear ion trap mass spectrometer. A single quadrupole offers Selected Ion Monitoring. A triple quadrupole adds Selected Reaction Monitoring. A linear ion trap goes further by enabling Consecutive Reaction Monitoring where a selected ion is fragmented into a product ion and the resulting product ion further fragmented in additional consecutive steps. Each additional fragmentation step makes compound identification more certain and facilitates structural characterization.
The LTQ XL ion trap MS extends the legendary MSn performance of the LTQ ion trap, incorporating more techniques to generate structural information through innovations in Thermo Scientific ion trap technology.
  • Unparalleled sensitivity and unmatched MSn spectral quality
  • Pulsed Q dissociation (PQD) for generating more fragments and extending the low mass range
  • High-resolution isolation (HRI) enables isolation of isobaric mass peaks for cleaner MSn spectra
  • Ultra-fast polarity switching for high sensitivity analysis of unknowns
  • Automated data-dependent neutral loss for PTM and biotransformation analyses
  • Excellent charge state determinations with Ultra ZoomScan™
The LTQ XL supplies outstanding sensitivity and abundant structural information for:
  • Protein identification and quantitative differential expression analysis
  • Biomarker studies and PTM identification
  • Metabolite identification and quantification
  • Tissue imaging
  • High-speed LC methods
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Amperage15 A
Frequency50/60 Hz
Polarity Switching<100 msec between positive and negative MS Scan Power
Scan RatesMSn for n = 1 through 10
SensitivityHeated Electrospray Ionization (HESI) - A Loop Injection of 2 μL of a 125 fg/μL Solution of Reserpine (250 Femtograms Total Sample). Will Produce a Minimum Signal-to-Noise Ratio of 100:1)
Voltage230 V
Weight (Metric)120 kg
Mass Range15 to 200 m/z, 50 to 2000 m/z, 200 to 4000 m/z
Resolution0.15 FWHM, 0.3 FWHM, 0.45 FWHM, 0.7 FWHM, 3 FWHM
TypeLTQ XL Linear Ion Trap MS
Unit SizeEach
Frequently asked questions (FAQs)
What are the indicators that I need to clean my Ion Trap mass spectrometry instrument?
If you notice poor sensitivity or the presence of large background ions, your instrument is in need of cleaning.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
How much of the Ion Trap mass spectrometry instrument can I clean?
You may clean the source housing, sweep cone, ion transfer tube, and the RF lens located behind the ITT.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
What tune file information is transferred to my Ion Trap mass spectrometry instrument method?
The tune file contains four items: Source parameters, ion optic parameters, AGC targets, and scan time settings.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
What does a proper cal mix look like on the Ion Trap mass spectrometry instruments, and what should my NL be?
See these charts for details.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
What are the differences between the various scan types on the Ion Trap mass spectrometry instruments?
See this table for details.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.