Paraformaldehyde Solution, 4% in PBS - FAQs
Paraformaldehyde Solution, 4% in PBS - FAQs
View additional product information for Paraformaldehyde Solution, 4% in PBS - FAQs (J19943.K2)
2 product FAQs found
Does 4% Paraformaldehyde in PBS contain methanol as a stabilizer?
No, 4% Paraformaldehyde in PBS does not contain methanol as a stabilizer.
At what temperature do you recommend storing 4% Paraformaldehyde in PBS (Cat. Nos. J61899.xx and J19943.K2)?
We recommend storing 4% Paraformaldehyde in PBS (Cat. Nos. J61899.xx and J19943.K2) at 2-8 degrees C.