Product Image
Thermo Scientific™

Maxima Probe/ROX qPCR Master Mix (2X)

**********************Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’veRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
K0231200 reactions
K02321,000 Reactions
K02334,000 Reactions
Catalog number K0231
Price (USD)
314.00
Each
No. of Reactions:
200 reactions
Price (USD)
314.00
Each
**********************
Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of Maxima Probe qPCR Master Mix and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific Maxima Probe qPCR Master Mixes are ready-to-use solutions optimized for quantitative real-time PCR. The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added.

Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG) (see Reference 1). The use of Maxima Probe qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates.

Maxima Probe qPCR Master Mixes are compatible with most real-time thermal cyclers.

Highlights

• Specificity—Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
• Sensitivity—detects low copy number targets
• Wide linear range—accurate quantification across 9 orders of magnitude
• Reproducibility and convenience—ready-to-use 2X master mix

Applications

• Gene expression
• siRNA validation
• Genotyping
• Pathogen detection

Includes
Maxima Probe/ROX qPCR Master Mix (2X) includes Maxima Hot Start Taq DNA Polymerase and dNTPs (also dUTP) in an optimized PCR buffer supplemented with ROX passive reference dye. Supplied with nuclease-free water.

Related Products
Maxima Probe/ROX qPCR Master Mix (2X)
Maxima Probe qPCR Master Mix (2X), with separate ROX vial
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
FormatTube
No. of Reactions200 reactions
PolymeraseTaq DNA Polymerase
Product LineMaxima
Product TypeqPCR Master Mix
Quantity200 reactions
Sample TypeDNA, dsDNA
Sufficient For200 Reactions of 25 μL
Volume2 x 1.25 mL
Concentration2X
Detection MethodPrimer-probe
For Use With (Application)Gene Expression
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Contains:
• 2 x 1.25 mL Maxima Probe/ROX qPCR Master Mix (2X) containing Maxima Hot Start Taq DNA Polymerase and dNTPs (also dUTP) in an optimized PCR buffer with ROX passive reference dye; sufficient for 200 x 25 μL reactions.
• 2 x 1.25 mL Nuclease-Free Water

Store at -20°C.