Product Image
Thermo Scientific™

Maxima Probe/ROX qPCR Master Mix (2X)

**********************Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호반응 수
K02321,000 Reactions
K0231200 reactions
K02334,000 Reactions
카탈로그 번호 K0232
제품 가격(KRW)
1,034,000
온라인 행사
Ends: 30-Jun-2026
1,148,000
할인액 114,000 (10%)
Each
반응 수:
1,000 Reactions
제품 가격(KRW)
1,034,000
온라인 행사
Ends: 30-Jun-2026
1,148,000
할인액 114,000 (10%)
Each
**********************
Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of Maxima Probe qPCR Master Mix and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific Maxima Probe qPCR Master Mixes are ready-to-use solutions optimized for quantitative real-time PCR. The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added.

Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG) (see Reference 1). The use of Maxima Probe qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates.

Maxima Probe qPCR Master Mixes are compatible with most real-time thermal cyclers.

Highlights

• Specificity—Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
• Sensitivity—detects low copy number targets
• Wide linear range—accurate quantification across 9 orders of magnitude
• Reproducibility and convenience—ready-to-use 2X master mix

Applications

• Gene expression
• siRNA validation
• Genotyping
• Pathogen detection

Includes
Maxima Probe/ROX qPCR Master Mix (2X) includes Maxima Hot Start Taq DNA Polymerase and dNTPs (also dUTP) in an optimized PCR buffer supplemented with ROX passive reference dye. Supplied with nuclease-free water.

Related Products
Maxima Probe/ROX qPCR Master Mix (2X)
Maxima Probe qPCR Master Mix (2X), with separate ROX vial
For Research Use Only. Not for use in diagnostic procedures.
사양
용도(장비)7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
형식Tube
반응 수1,000 Reactions
중합효소Taq DNA Polymerase
제품라인Maxima
제품 유형qPCR Master Mix
수량1,000 reactions
샘플 종류DNA, dsDNA
충분1000 Reactions at 25 μL
부피10 x 1.25 mL
농도2X
검출 방법Primer-probe
용도(애플리케이션)Gene Expression
GC-Rich PCR PerformanceHigh
PCR 방법qPCR
반응 속도Standard
Unit SizeEach
구성 및 보관
Contains:
• 10 x 1.25 mL Maxima Probe/ROX qPCR Master Mix (2X) containing Maxima Hot Start Taq DNA Polymerase and dNTPs (also dUTP) in an optimized PCR buffer with ROX passive reference dye; sufficient for 1000 x 25 μL reactions.
• 10 x 1.25 mL Nuclease-Free Water

Store at -20°C.